Search
Search

| Catalog Number | Quantity | For Use With (Equipment) |
|---|---|---|
| 900062 | 5 x 60 mL | Chloride ISE, Cyanide ISE, Lead ISE, Silver/Sulfide ISE |
| 900010 | 5 x 60 mL | Sodium ISE |
| 900012 | 5 x 60 mL | Sodium ISE (Low Sodium) |
| 900017 | 5 x 60 mL | Chloride ISE |
| 900018-WA | 5 x 60 mL | Ammonium ISE |
| 900046 | 5 x 60 mL | Nitrate ISE |
| 900061 | 5 x 60 mL | Cadmium ISE, Calcium ISE, Fluoride ISE, Sulfide ISE |
| 900063 | 5 x 60 mL | Bromide ISE, Cupric ISE, Iodide ISE |
| 900065 | 5 x 60 mL | Potassium ISE |
| 951202 | 60 mL | Ammonia ISE |
| 951209 | 60 mL | Ammonia ISE |
Enhance the performance and extend the life of your ion selective electrodes with Thermo Scientific™ Orion™ ISE Filling Solutions. These solutions are specially formulated to work with the recommended ion selective electrodes and prevent electrode interferences from filling solution.
Ordering Information:
Note:
The following part number has been changed due to a recent system upgrade:
Old Part Number: 900018
New Part Number: 900018-WA
Please use the new part number when placing an order or requesting a quote. The old and new part numbers are interchangeable on Certificate of Analysis and Material Safety Data Sheet (MSDS) files.
Warranty: Out of box