GeneChip™ Test3 Array
Applied Biosystems™
GeneChip™ Test3 Array
The GeneChip™ Test3 Array provides a convenient and accurate method to determine the quality of a labeled target prior toWeitere Informationen
Have Questions?
KatalognummerMenge
90034230 Arrays
Katalognummer 900342
Preis (EUR)
-
Menge:
30 Arrays
The GeneChip™ Test3 Array provides a convenient and accurate method to determine the quality of a labeled target prior to its analysis the GeneChip expression arrays.
The GeneChip Test3 Array provides a convenient and accurate method to determine the quality of a labeled target prior to its analysis the GeneChip expression arrays. This array contains probe sets representing a subset of characterized genes from various organisms, including mammals, plants, and eubacteria.
Additionally, the GeneChip Test3 Array contains a subset of human and mouse housekeeping genes (shown to be expressed early in fetal development and throughout adulthood). This array also serves as an economically priced training tool for new GeneChip technology users.
For more information, please review the data sheet (pdf, 132 KB).
Nur für Forschungszwecke. Nicht zur Verwendung bei diagnostischen Verfahren.
Specifications
Menge30 Arrays
TypTest3 Array
ArrayTranscriptome Profiling
FormatArray Cartridge
Anzahl Arrays30 arrays
SpeziesAll Organisms
Unit SizeEach