GeneChip™ T7-Oligo(dT) Promoter Primer Kit
Product Image
Applied Biosystems™
GeneChip™ T7-Oligo(dT) Promoter Primer Kit
This reagent kit is an essential component of the eukaryotic target labeling procedure for expression analysis using GeneChip™ probe arrays.Read more
Have Questions?
Catalog NumberQuantity
9003751 kit
Catalog number 900375
Price (USD)
-
Quantity:
1 kit
This reagent kit is an essential component of the eukaryotic target labeling procedure for expression analysis using GeneChip™ probe arrays. It contains 50 μM T7-Oligo(dT) promoter primer and RNase-free water.
Each T7-Oligo(dT) Promoter Primer Kit provides sufficient primer for 150 labeling reactions. All components are ready to be added directly to RNA samples to initiate the first strand cDNA synthesis reaction. This reagent kit has been tested, is quality assured, and is able to sustain at least twenty-five freeze-thaw cycles without compromise to its performance.
For more information, please review the data sheet (152 KB).
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Product TypeOligonucleotides, Primers
Quantity1 kit
PrimerT7
Product LineGeneChip
PromoterT7
Unit SizeEach