Product Image
Applied Biosystems™
GeneChip™ C. elegans Genome Array
This whole genome array represents 22,500 transcripts from Caenorhabditis elegans, a model organism for developmental biology and functional genomics.
Have Questions?
Catalog NumberQuantity
9003835 arrays
Catalog number 900383
Price (USD)
-
Quantity:
5 arrays
This whole genome array represents 22,500 transcripts from Caenorhabditis elegans, a model organism for developmental biology and functional genomics.
Sequence information used to select these probe sets came from the December 05, 2000 Sanger Center database release and GenBank™ release 121. All sequences were functionally re-annotated by us for improved descriptions.
Related Links:
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Product LineGeneChip
Quantity5 arrays
TypeC. elegans Genome Array
ArrayTranscriptome Profiling
FormatArray Cartridge
Number of Arrays5 arrays
SpeciesC. elegans
Unit Size5 arrays