Product Image
Applied Biosystems™

Attune™ Flow Cytometer Shutdown Bottle

This is a 250 mL replacement bottle for shutdown solution for the Attune Flow Cytometer.
Have Questions?
Catalog NumberQuantity
900392741 each
Catalog number 90039274
Price (MXN)
-
Quantity:
1 each

This is a 250 mL replacement bottle for shutdown solution for the Attune Flow Cytometer.

For Research Use Only. Not for use in diagnostic procedures.

Specifications
Product LineAttune
Quantity1 each
Shipping ConditionRoom Temperature
For Use With (Equipment)Attune Acoustic Focusing Cytometer
Product TypeShutdown Bottle
Unit SizeEach
Contents & Storage
Store at room temperature