GeneChip™ Hybridization Control Kit - FAQs
GeneChip™ Hybridization Control Kit - FAQs
View additional product information for GeneChip™ Hybridization Control Kit - FAQs (900457)
3 product FAQs found
How many freeze/thaws has the 20X Hybridization Control Stock, in the GeneChip Hybridization Control Kit, been validated for?
The 20X Hybridization Control Stock has been validated for 3 freeze/thaws.
Find additional tips, troubleshooting help, and resources within our Microarray Analysis Support Center.
What are the contents of the GeneChip Hybridization Control Kits?
We offer two GeneChip Hybridization Control Kits, that differ in size:
- Cat. No. 900454: 30 rxns
- Cat. No. 900457: 150 rxns
Both contain 20X Hybridization Control Shock and Control Oligo B2.
Find additional tips, troubleshooting help, and resources within our Microarray Analysis Support Center.
Should I always anticipate the hybridization controls, bioB, bioC, bioD, and cre, to be called as Present?
The four transcripts are added to the hybridization cocktail at staggered concentrations. At 1.5 pM, bioB is at the detection limit for most expression arrays and is anticipated to be called Present at least 50% of the time. In contrast, the other controls should be called Present all of the time, with increasing Signal values (bioC, bioD, and cre, respectively).
Find additional tips, troubleshooting help, and resources within our Microarray Analysis Support Center.