GeneChip™ Human Genome U133 Plus 2.0 Array, 30 arrays - FAQs
GeneChip™ Human Genome U133 Plus 2.0 Array, 30 arrays - FAQs
View additional product information for GeneChip™ Human Genome U133 Plus 2.0 Array - FAQs (900467, 900466)
10 product FAQs found
How are transcript IDs assigned for WT arrays if a gene has multiple transcript IDs?
The first assigned mRNA/gene that is listed in the annotation file for a given probe set or transcript cluster is considered the "best" assignment based on data source quality ranking and assignment scoring system. To get a single best mRNA/gene assignment you can simply take the first one in the list in the annotation file.
Sometimes, there might be multiple best hits between transcript clusters (or probe set) and a mRNA/gene, with identical score and data source quality. In that case, the transcript assignments are ranked based on their descriptions (e.g., title from the GenBank record).
For transcript clusters that detect genes that are alternatively spliced, the different spliced forms will be ranked by length with the longest ones listed first.
Find additional tips, troubleshooting help, and resources within our Microarray Analysis Support Center.
Does the GeneChip miRNA 3.1 Array Strip require a different hyb/wash/stain kit than the GeneChip 3' IVT arrays and GeneChip ST array strips?
The hyb/wash/stain kit for the GeneChip miRNA 3.1 Array Strips is different than that for the 3' IVT arrays and GeneChip ST array strips. Additional wash A steps have been added to ensure adequate washing after the array strips have been stained.
Find additional tips, troubleshooting help, and resources within our Microarray Analysis Support Center.
What do the suffixes mean at the end of each probeset in GeneChip 3' IVT arrays?
The suffixes, at the end of each probeset in GeneChip 3' IVT arrays, indicate what is known about the sequence at the time of design:
_at = all the probes hit one known transcript
_a = all probes in the set hit alternate transcripts from the same gene
_s = all probes in the set hit transcripts from the different genes
_x = some probes hit transcripts from different genes
Find additional tips, troubleshooting help, and resources within our Microarray Analysis Support Center.
Can I store my GeneChip 3' IVT Array after hybridization?
Yes, but we recommend storing your GeneChip 3' IVT Array after hybridization and prior to the fluidics step, for no longer than 3 hours at 4 degrees C. Please remove the hybridization cocktail and fill with wash A before storing.
Find additional tips, troubleshooting help, and resources within our Microarray Analysis Support Center.
What does the "_x_at" extension represent in the HG-U133 probe set name?
Occasionally, it is not possible to select either a unique probe set or a probe set with all probes common among multiple transcripts ("_s_at" ). In such cases, similarity criteria are suspended, and the resulting probe set name is appended with the "_x_at" extension. These probe sets contain some probes that are identical, or highly similar, to unrelated sequences. These probes may cross-hybridize in an unpredictable manner with sequences other than the main target. Data generated from these probe sets should be interpreted with caution, due to the likelihood that some of the signal is from transcripts other than the one being intentionally measured.
Find additional tips, troubleshooting help, and resources within our Microarray Analysis Support Center.
What does the "_s_at" extension represent in the HG-U133 probe set name?
The primary goal in probe set selection is to select a probe set unique to a single transcript or common among a small set of similar transcript variants. A probe set name is appended with the "_s_at" extension when all the probes exactly match multiple transcripts. The probe set selection process generally favors probe sets measuring fewer transcripts. Probe sets with common probes among multiple transcripts (the "_s_at" probe sets), are frequent and are to be expected, due to alternative polyadenylation and alternative splicing. In most cases, "_s_at" probe sets represent transcripts from the same gene, but the same probe set can sometimes also represent transcripts from homologous genes. One transcript may be represented by both a unique and an "_s_at" probe set when the transcript variation is sufficient.
Find additional tips, troubleshooting help, and resources within our Microarray Analysis Support Center.
How is this array design principle different from the GeneChip Human Genome U133 type of arrays?
The GeneChip Exon Arrays represent a new array design philosophy for the most comprehensive and informative coverage with the exon as the basic unit of expression analysis. Such an inclusive design enables discovery of new and novel splicing events not previously observed experimentally. Some of the key differences are summarized below using the human arrays as an example; refer to the “GeneChip Exon Array Design” Technical Note and the “Design and Performance of the GeneChip Human Genome U133 Plus 2.0 and Human Genome U133A 2.0 Array” Technical Note for more details.
Find additional tips, troubleshooting help, and resources within our Microarray Analysis Support Center.
On average, how many probes are used to represent a corresponding GeneChip Human Genome U133 (HG-U133) Array probe set where there are 11 probe pairs?
The majority of the HG-U133 Plus 2.0 Array consensus sequences are well covered in the new design with approximately 75% of them containing over 8 probes on the Exon Array. The remaining 25% have less overlap with the new exon probes, and are mostly attributed to EST-based HG-U133 Array probe sets that do not contain an annotated coding region. As a result, in many such cases, the entire HG-U133 Array EST-based consensus sequence is designated as one PSR, with around 4 probes selected for that entire region.
Find additional tips, troubleshooting help, and resources within our Microarray Analysis Support Center.
Can I compare Exon Array results with those obtained on the GeneChip Human Genome U133 (HG-U133) Array?
Signal and detection values from the exon arrays cannot be directly compared to that of the HG-U133 arrays. Major differences in array design and assay prevent meaningful comparisons at the signal level. Splice variation and polyadenylation variation can confound comparisons at the biological level (i.e., direction of change) due to differences in probe placement and bias in the target preparation assays.
Find additional tips, troubleshooting help, and resources within our Microarray Analysis Support Center.