GeneChip™ Vitis vinifera (Grape) Genome Array
Applied Biosystems™
GeneChip™ Vitis vinifera (Grape) Genome Array
The GeneChip™ Vitis vinifera Genome Array is the first array to provide comprehensive coverage of the V. vinifera genome.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberNumber of Arrays
9005092 arrays
9005106 arrays
Catalog number 900509
Price (USD)
-
Number of Arrays:
2 arrays
The GeneChip™ Vitis vinifera Genome Array is the first array to provide comprehensive coverage of the V. vinifera genome.
Convenient one-array view
  • 14,000 V. vinifera transcripts and 1,700 transcripts from other Vitis species can be interrogated
  • Use the Power of the Probe Set and get multiple independent measurements for each transcript that deliver the greatest accuracy and reproducibility of any microarray platform
  • GeneChip System: Richer data, sharper insights, better decisions
Array Profile
Oligonucleotide probes complementary to each corresponding sequence are synthesized in situ on the arrays. Sixteen pairs of oligonucleotide probes are used to measure the level of transcription of each sequence represented on the GeneChip Vitis vinifera (Grape) Genome Array. Sequences used in the design of the V. vinifera Genome Array were selected from GenBank™, dbEST, and RefSeq. The sequence clusters were created from the UniGene database (Build 7, Oct 2003).
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Product LineGeneChip
Quantity2 arrays
TypeVitis vinifera (Grape) Genome Array
ArrayTranscriptome Profiling
FormatArray Cartridge
Number of Arrays2 arrays
SpeciesVitis vinifera (Grape)
Unit Size2 arrays