Applied Biosystems™

GeneChip™ E. coli Genome 2.0 Array

Catalog number: 900550
Applied Biosystems™

GeneChip™ E. coli Genome 2.0 Array

Catalog number: 900550
Catalog Number
900550
Unit Size
2 arrays
Price (USD)
Price: 952.00
Your Price:
Availability
-
Quantity
Catalog Number
Unit Size
6 arrays
Price (USD)
Price: 2,730.00
Your Price:
Availability
-
Quantity
Catalog Number
Unit Size
30 arrays
Price (USD)
Price: 14,070.00
Your Price:
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
9005502 arrays
Price: 952.00
Your Price:
-
9005516 arrays
Price: 2,730.00
Your Price:
-
90055230 arrays
Price: 14,070.00
Your Price:
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
The GeneChip™ E. coli Genome 2.0 Array contains probe sets to detect transcripts from the K12 strain of E. coli and three pathogenic strains of E. coli. Using the E. coli Genome 2.0 Array, researchers can gain a comprehensive view of three pathogenic forms of E. coli while continuing ongoing research projects with the K12 strain.

The GeneChip E. coli Genome 2.0 Array includes approximately 10,000 probe sets for all 20,366 genes present in four strains of E. coli. Due to the high degree of similarity between the E. coli strains, whenever possible, a single probe set is tiled to represent the equivalent ortholog in all four strains. The GeneChip E. coli Genome 2.0 Array tiles probe sets over the entire open reading frame (ORF) of E. coli, includes over 700 intergenic regions, probes for various antibiotic resistance markers, and incorporates additional control and reporter genes from the previous generation E. coli array.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Type
E. coli Genome 2.0 Array
Array
Transcriptome Profiling
Number of Arrays
2 arrays
Format
Array Cartridge
Species
E. coli
Product Line
GeneChip™

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text