GeneChip™ Rice Genome Array
GeneChip™ Rice Genome Array
Applied Biosystems™
GeneChip™ Rice Genome Array
The GeneChip™ Rice Genome Array offers researchers comprehensive transcriptome coverage for rice gene expression research. This array contains probes toRead more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberNumber of Arrays
9006006 arrays
9005992 arrays
90060130 arrays
Catalog number 900600
Price (MXN)
-
Number of Arrays:
6 arrays
The GeneChip™ Rice Genome Array offers researchers comprehensive transcriptome coverage for rice gene expression research. This array contains probes to query 51,279 transcripts representing two rice cultivars, with approximately 48,564 japonica transcripts and 1,260 transcripts representing the indica cultivar. This unique design was created within the GeneChip Consortia Program and provides scientists with a single array that can be used for the study of rice. High-quality sequence data were derived from GenBank™ mRNAs, TIGR gene predictions, and the International Rice Genome sequencing project. The arrays were designed using NCBI UniGene Build #52, (May 7, 2004) incorporating predicted genes from GenBank and the TIGR Os1 v2 data set. (ftp.tigr.org FASTA, 89.3 MB).
Instrument and Software Requirements
• GeneChip Scanner 3000, enabled for High-Resolution Scanning*
• GeneChip Operating Software (GCOS) v1.1.1 or higher, which includes the GeneChip Scanner 3000 High-Resolution Scanning Patch
* GeneChip Scanner 3000 High-Resolution Update is standard on all instruments shipped starting in September 2003 with serial number series 502. Previous versions, such as serial number series 501, will require the GeneChip Scanner 3000 High-Resolution Update, Catalog Number 00-0110, to be installed.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Product LineGeneChip
Quantity6 arrays
TypeRice Genome Array
ArrayTranscriptome Profiling
FormatArray Cartridge
Number of Arrays6 arrays
SpeciesRice
Unit Size6 arrays