GeneChip™ S. cerevisiae Tiling 1.0R Array
Applied Biosystems™
GeneChip™ S. cerevisiae Tiling 1.0R Array
The GeneChip™ S. cerevisiae Tiling 1.0R Array is a versatile whole-genome research tool:• Most comprehensive S. cerevisiae microarray offering high-densityRead more
Have Questions?
Catalog NumberQuantity
9006456 arrays
Catalog number 900645
Price (USD)
7,240.00
6 arrays
Quantity:
6 arrays
Price (USD)
7,240.00
6 arrays
The GeneChip™ S. cerevisiae Tiling 1.0R Array is a versatile whole-genome research tool:
• Most comprehensive S. cerevisiae microarray offering high-density 5 base pair probe resolution
• High-density array design is compatible with many research applications
• 25-mer probes yield the most specific hybridization results available
Array Profile
The GeneChip S. cerevisiae Tiling 1.0R Array is designed for identifying novel transcripts, mapping sites of protein/DNA interaction in chromatin immunoprecipitation (ChIP) experiments, and other whole-genome experiments.* The S. cerevisiae 1.0R Array is a single array comprised of over 3.2 million perfect match/mismatch probe pairs tiled through the complete Saccharomyces cerevisiae genome. Genome sequence information for the design of the S. cerevisiae Tiling 1.0R Array was drawn from the October 2003 Stanford Yeast Genome Database files (www.yeastgenome.org). The array also contains probes to interrogate a 2-μm circle plasmid (NCBI Accession J01347).
Probes are tiled at an average of 5 base pair resolution, as measured from the central position of adjacent 25-mer oligos, creating an overlap of approximately 20 base pairs on adjacent probes.
*Gresham D, et al. Genome-wide detection of polymorphisms at nucleotide resolution with a single DNA microarray. Science 311(5769):1932-6 (2006 Mar 31).
Instrument and Software Requirements
• GeneChip Scanner 3000 7G
• GeneChip Operating Software (GCOS) version 1.3 or higher
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Product LineGeneChip
Quantity6 arrays
TypeS. cerevisiae Tiling 1.0R Array
ArrayTranscriptome Profiling
FormatArray Cartridge
Number of Arrays6 arrays
SpeciesS. cerevisiae
Unit Size6 arrays
Frequently asked questions (FAQs)
Do you offer any annotation files for the GeneChip S. cerevisiae Tiling 1.0R Array?
We do not provide annotation files for the GeneChip S. cerevisiae Tiling 1.0R Array, but we do provide annotation files for human and mouse promoter arrays.
Find additional tips, troubleshooting help, and resources within our Microarray Analysis Support Center.
Is the GeneChip S.cerevisiae Tiling 1.0R Array designed from the forward strand or the reverse strand?
In the name of this product, GeneChip S. cerevisiae Tiling 1.0R Array, the 'R' indicates that the probe was designed from the reverse strand.
Find additional tips, troubleshooting help, and resources within our Microarray Analysis Support Center.