TMTzero™ Label Reagent, 5 x 0.8 mg
Thermo Scientific™

TMTzero™ Label Reagent, 5 x 0.8 mg

The Thermo Scientific™ TMTzero Label Reagent serves as a 'light' tag for MS-level peptide quantitation similar to duplex isotopic metabolic labeling, such as SILAC, or isotopic dimethylation labeling.
Have Questions?
Catalog NumberQuantity
900675 x 0.8 mg
Catalog number 90067
Price (KRW)
373,000
Online offer
Ends: 31-Dec-2026
426,000
Save 53,000 (12%)
Each
Quantity:
5 x 0.8 mg
Request bulk or custom format
Price (KRW)
373,000
Online offer
Ends: 31-Dec-2026
426,000
Save 53,000 (12%)
Each

The amine-reactive, NHS-ester-activated compounds covalently attach to the peptide amino terminus and free amino termini of lysine residues of peptides and proteins with high efficiency, thereby labeling all peptides in each sample regardless of enzyme used for digestion. Since TMT reagents share an identical structure and mass (i.e., isotopomers), labeled peptides co-elute during LC separation and are co-isolated during MS/MS analysis, resulting in fewer missing peptide identifications among samples. During MS/MS analysis, each isobaric tag is also fragmented to produce a unique reporter ion mass that is used for sample identification and quantitation. Protein quantitation is accomplished by comparing the relative intensities of the two reporter ions in the MS/MS spectra.

For Research Use Only. Not for use in diagnostic procedures.

Specifications
DescriptionPierce TMTzero Label Reagent, 5 x 0.8 mg
Final Product TypeProtein
For Use With (Equipment)Mass Spectrometer
Label or DyeTandem Mass Tag™ (TMT) Labels
Quantity5 x 0.8 mg
Workflow StepProtein Labeling
Detection MethodMass Spectrometry
Product LineTMTzero
Starting MaterialCell Lysate
Unit SizeEach
Contents & Storage
Store at –20°C.

Frequently asked questions (FAQs)

Can the TMT and TMTpro reagents be mixed?

No, the TMT and TMTpro reagents cannot be mixed because the reporter ions are in the same region.

Find additional tips, troubleshooting help, and resources within our Mass Spectrometry Support Center.