900804 - FAQs
900804 - FAQs
View additional product information for 900804 - FAQs (900804)
4 product FAQs found
How long does it take to scan a Mapping 500K array?
Mapping 500K array scans require ~30 minutes per array.
Find additional tips, troubleshooting help, and resources within our Microarray Analysis Support Center.
How many SNPs and Copy Number Probes are on the Mapping 500K array set?
There are approximately 262,000 NSP SNPs and approximately 238,000 STY SNPs that are used for SNP and copy number analysis.
Find additional tips, troubleshooting help, and resources within our Microarray Analysis Support Center.
At what temperature should customers keep the cooling chamber for the 500K protocol?
Keep the cooling chamber at 4 degrees C. NEVER freeze at -20 degrees C because ice can form in the wells.
Find additional tips, troubleshooting help, and resources within our Microarray Analysis Support Center.
When should 500K enzymes be vortexed?
The enzyme alone should never be vortexed. The final cocktail, when specified, can be vortexed.
Find additional tips, troubleshooting help, and resources within our Microarray Analysis Support Center.