GeneChip™ Human Gene 1.0 ST Array
Applied Biosystems™

GeneChip™ Human Gene 1.0 ST Array

The GeneChip™ Human Gene 1.0 ST Array provides the most accurate, sensitive, and comprehensive measurement of protein coding and long intergenic non-coding RNA transcripts.
Have Questions?
Cambiar vistabuttonViewtableView
Número de catálogoNúmero de arrays
9010866 matrices
9010852 matrices
90108730 matrices
Número de catálogo 901086
Precio (MXN)
-
Número de arrays:
6 matrices

The GeneChip™ Human Gene 1.0 ST Array provides the most accurate, sensitive, and comprehensive measurement of protein coding and long intergenic non-coding RNA transcripts. We also offer the GeneChip Human Gene 2.0 ST Array, which can measure differential expression of long intergenic non-coding RNA transcripts.

For Research Use Only. Not for use in diagnostic procedures.

Especificaciones
Línea de productosGeneChip
Cantidad6 arrays
TipoHuman Gene 1.0 ST Array
matriz, red, conjuntoTranscriptome Profiling
FormatoArray Cartridge
Número de arrays6 matrices
EspecieHumano
Unit Size6 arrays

Preguntas frecuentes

What is contained in the tab-delimited format of the GeneChip probe sequence download file?

The tab-delimited probe sequence file contains the following information:
-Probe Set Name
-Probe X: The X coordinate of the probe sequence on the GeneChip probe array.
-Probe Y: The Y coordinate of the probe sequence on the GeneChip probe array.
-Probe Interrogation Position: The base position on the consensus/exemplar sequence where the central base of the probe aligns, which is the 13th base of a 25mer probe.
-Probe Sequence: The 25-base perfect match sequence.
-Target Strandedness: The sense/antisense orientation of the target sequence that can hybridize with the probe sequence.

Find additional tips, troubleshooting help, and resources within our Microarray Analysis Support Center.

What is the NetAffx Analysis Center?

The NetAffx Analysis Center used to contain information and files regarding array content, probe sets, and functional annotations. However, the NetAffx Analysis Center has been retired. Much of the content provided by the NetAffx Analysis Center can now be accessed on the arrays’ thermofisher.com product pages or by contacting Technical Support (techsupport@thermofisher.com).

Find additional tips, troubleshooting help, and resources within our Microarray Analysis Support Center.

What reagent kit should I use with my array?

Please refer to the Microarray Reagent Guide for Arrays and Expression Kits to match the correct reagents your array.

Find additional tips, troubleshooting help, and resources within our Microarray Analysis Support Center.

Are pseudogene databases included in the design of expression arrays?

Pseudogene databases were not included in the design of expression arrays.

How long can I store labeled cDNA when working with expression microarrays?

Labeled material can be stored for 2 weeks at -20 degrees C.