Applied Biosystems™
GeneAtlas™ Hybridization, Wash, and Stain Kit for WT Array Strips
Catalog number:  901667
Related applications:
Microarray Analysis
Back to top
Description
The GeneAtlas™ Hybridization, Wash, and Stain Kit for WT Array Strips contains pre-formulated solutions to process whole-transcript (WT) array strips using the GeneAtlas System. The reagents are formulated into the fewest number of individual components possible, reducing preparation and handling steps. With enhanced performance, fewer required reagents, and a simple protocol, target preparation has never been so easy.
The GeneAtlas Hybridization, Wash, and Stain Kit for WT Array Strips includes two boxes. The first box contains sufficient amounts of the hybridization and stain components to process 15 array strips (60 arrays). The hybridization components include the solutions required for preparation of the hybridization cocktail containing the target. The stain components include Stain Cocktail 1 and 3, Stain Cocktail 2, and the Array Holding Buffer. These solutions are used during array processing using the GeneAtlas System.
The second box includes Wash Buffer A and Wash Buffer B, also required for processing arrays. There is sufficient Wash Buffer A and B to process 15 array strips (60 arrays).
The Stain Cocktail 1 and 3, Stain Cocktail 2, Array Holding Buffer, and Wash Buffers A and B are ready-to-use solutions. The hybridization cocktail is prepared by combining the Hybridization Mix and nuclease-free water with the target prepared with the WT target labeling assay.
The GeneAtlas Hybridization, Wash, and Stain Kit for WT Array Strips should always be stored at 2 to 8°C.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
For Use With (Application): Transcriptome Profiling
Product Size: 1 kit
Reagent Type: Hybridization, Wash, Stain