Axiom™ Genome-Wide EUR, 96 array plate
Applied Biosystems™

Axiom™ Genome-Wide EUR, 96 array plate

Axiom™ Genome-Wide EUR 1 Array Plate is designed for European ancestry genome-wide imputation analysis with a focus in disease-associated genes.Read more
Have Questions?
Catalog NumberQuantity
9017871 plate
Catalog number 901787
Price (USD)
-
Quantity:
1 plate
Axiom™ Genome-Wide EUR 1 Array Plate is designed for European ancestry genome-wide imputation analysis with a focus in disease-associated genes. The arrays include additional markers that are specific to multiple HapMap populations. These arrays have the industry-s highest coverage of rare variants to replicate studies of specific loci and/or genes that have been identified in previous genome-wide association studies. The use of imputation enables a highly cost-effective, budget conscious alternative to other commercially available arrays.

We also offer the following Maximum Genome-Wide Coverage Arrays:

• Axiom Genome-Wide ASI 1 Array Plate - The first array designed to maximize genomic coverage of rare alleles (MAF >1%) of a consensus East Asian (JPT+CHB) genome

• Axiom Genome-Wide CHB 1 & CHB 2 Array Plate - The Genome-Wide CHB 1 Array Plate maximizes genomic coverage of common alleles (MAF >5%) of the Han Chinese genome. The Genome-Wide CHB 2 Array Plate is designed to target rare variants (MAF 2-5%) as a complement to Axiom Genome-Wide CHB 1 Array Plate. The Genome-Wide CHB 1 & CHB 2 Array Set Bundle enables the most comprehensive commercially available genome-wide coverage in CHB populations.

• Axiom Genome-Wide PanAFR Array Plate Set - The first array to offer pan-African genomic coverage, with ˜90% genetic coverage of common and rare variants (MAF >2%) of the Yoruba (West African) genome and >85% coverage of common and rare variants (MAF >2%) of the Luhya and Maasi (East African) genomes. This array set also offers high genomic coverage (>85%) in admixed populations with West African ancestry.

• Axiom Genome-Wide CEU 1 Array Plate Our first cost-effective array to maximize coverage of rare variants (MAF >1%) for European populations.

• Axiom Genome-Wide EAS 1 Array Plate - East Asian ancestry

• Axiom Genome-Wide AFR 1 Array Plate - Admixed population of West African and European ancestry

• Axiom Genome-Wide LAT 1 Array Plate - Admixed population of Native American, European, and West African ancestry

Don't see what you-re looking for? Please contact us for assistance in choosing a product or to request an array design consulatation.

Benefits of Axiom Genome-Wide Population-Optimized Arrays:

Each array plate was designed to achieve ˜90% genomic coverage (r2 >0.8) in each target population, while covering markers with MAF of 1-5% in areas such as:
• Critical biological categories (e.g., coding SNPs)
• Biological processes (e.g., drug metabolizing genes)
• Disease categories (e.g., cardiovascular disease, cancer, immunity/inflammation, MHC, and CNS)
• Common and rare SNPs and insertions/deletions (indels) from the International HapMap Project, 1000 Genomes Project, and published disease associations
• Genotype-tested genomic content proven to give informative and reliable results
• Markers preselected for coverage in critical biological categories, such as coding SNPs, and biological pathways, such as drug metabolizing or cardiovascular genes
• High sample pass rate, call rate, and reproducibility
• Compatible with the Axiom 2.0 Reagent Kit, GeneTitan™ MC Instrument, automated or manual workflow, and Genotyping Console™ Software
For Research Use Only. Not for use in diagnostic procedures.
Specifications
For Use With (Application)Microarray Analysis
Quantity1 plate
ArrayGenotyping
Format96-array Plate
Number of Arrays96 arrays
SpeciesHuman
Unit SizeEach

Frequently asked questions (FAQs)

Is the hybridization mix for cartridges the same as the hybridization mix for PEG arrays (array plates)?

The hybridization mix for PEG arrays is different from the hybridization mix for cartridges. The concentrations of the hybridization mix are different and in addition, DMSO is not added into the hybridization mix for PEG arrays because it can affect the glue that holds the PEG to the plate.

What is the starting amount of DNA required for Axiom arrays?

All Axiom arrays (except for Axiom Genome-Wide Pan-African Array Set) require a total of 200 ng DNA. The Axiom Genome-Wide Pan-African Array Set requires a total of 300 ng DNA, or 100 ng DNA per array (there are 3 arrays).

Find additional tips, troubleshooting help, and resources within our Microarray Analysis Support Center.

What are the sample requirements for Axiom array processing at the Thermo Fisher Scientific Microarray Research Services Laboratory (MRSL)?

For information regarding our in-house MRSL requirements, please contact us via phone at 888-362-2447 (in North America) or via email at BioinformaticsServices@thermofisher.com.

Find additional tips, troubleshooting help, and resources within our Microarray Analysis Support Center.