Axiom™ Genome-Wide Chicken Array Kit
Applied Biosystems™
Axiom™ Genome-Wide Chicken Array Kit
Axiom Chicken Genotyping Array was developed in collaboration with leading academic institutions and commercial poultry companies including the Roslin Institute,Más información
Have Questions?
Número de catálogoCantidad
9021481 kit
Número de catálogo 902148
Precio (MXN)
-
Cantidad:
1 kit
Axiom Chicken Genotyping Array was developed in collaboration with leading academic institutions and commercial poultry companies including the Roslin Institute, Aviagen Ltd, Hy-Line International under BBSRC LINK funding and the German Synbreed project funded by the BMBF.
This genotyping array detects genetic variants in white egg layers, brown egg layers, broilers and outbred non-commercial breeds. The parental high-density information from the array is useful for poultry breeding while markers associated with wild out-bred lines are useful for analysis of genetic diversity among indigenous chicken populations.
Features and benefits of Axiom Chicken Genotyping Array:
Comprehensive content: Over 580,000 common and rare markers selected from 1.8M tested variants
Genome-wide coverage: Well-annotated markers uniformly distributed across the genome in layers and broilers
Designed for your breed: Over 24 breeds represented including commercial layers and broilers and non-commercial outbred lines
Affordable: High return on investment and lowest available cost per marker
Robust assay performance: Includes genotype-tested SNPs that are highly polymorphic and demonstrate a >99% call rate
Applications of Axiom Chicken Genotyping Array:
Genome-wide association studies
High-resolution genetic mapping
Mendelian trait mapping
Diversity analysis in indigenous chickens
Selection signature analysis
Required Products
Axiom Analysis Suite
GeneChip Command Console Software
GeneTitan Multi-Channel Instrument
Para uso exclusivo en investigación. No apto para uso en procedimientos diagnósticos.
Especificaciones
Para utilizar con (aplicación)Microarray Analysis
Cantidad1 kit
TipoGenome-Wide Chicken Array Kit
matriz, red, conjuntoGenotyping
Formato96-array Plate
Número de arrays96 matrices
EspecieChicken
Unit SizeEach
Preguntas frecuentes
What is the starting amount of DNA required for Axiom arrays?
All Axiom arrays (except for Axiom Genome-Wide Pan-African Array Set) require a total of 200 ng DNA. The Axiom Genome-Wide Pan-African Array Set requires a total of 300 ng DNA, or 100 ng DNA per array (there are 3 arrays).
Find additional tips, troubleshooting help, and resources within our Microarray Analysis Support Center.
What are the sample requirements for Axiom array processing at the Thermo Fisher Scientific Microarray Research Services Laboratory (MRSL)?
For information regarding our in-house MRSL requirements, please contact us via phone at 888-362-2447 (in North America) or via email at BioinformaticsServices@thermofisher.com.
Find additional tips, troubleshooting help, and resources within our Microarray Analysis Support Center.