Product Image
Applied Biosystems™
Axiom™ 2.0 384HT Reagent Kit
Axiom™ 2.0 384HT Reagent Kit includes all reagents to process one Axiom 384HT Array Plate.
Have Questions?
Número de catálogoCantidad
9022451 placa
Número de catálogo 902245
Precio (CLP)
-
Cantidad:
1 placa
Axiom™ 2.0 384HT Reagent Kit includes all reagents to process one Axiom 384HT Array Plate.
Para uso exclusivo en investigación. No apto para uso en procedimientos diagnósticos.
Especificaciones
Cantidad1 placa
Tipo de reactivoGenotyping reagents
TipoReagent Kit
Unit SizeEach
Preguntas frecuentes
Why do I need to use the Control Oligo B2 with my assay?
Control Oligo B2 (3 nM) is a pre-labeled DNA spike-in control required for hybridization to the control probes on the array utilized for grid alignment by AGCC. A failure to include Control Oligo B2 in the hybridization cocktail will result in the inability of the software to apply a grid over the scanned image, leading to the unrecoverable loss of sample data from the array. The absence of the Control Oligo B2 also voids the customer's ability to submit an array replacement request.
Find additional tips, troubleshooting help, and resources within our Microarray Analysis Support Center.