Search
| Catalog Number | Quantity |
|---|---|
| 902288 | 1 kit |
The Axiom™ 384HT Consumables Kit for Biomek™ FXp is required for target preparation on the Biomek FXp and contains enough reagents for five Axiom 384-Array Plates.
Based on the universal workflow of the Genotyping Solution, the myDesign Arrays offer unprecedented scale and flexibility to design the most optimal array for your species of interest.
For Research Use Only. Not for use in diagnostic procedures.