Product Image
Applied Biosystems™
GeneChip™ 3' IVT PLUS Reagent Kit
The GeneChip™ 3' IVT PLUS Reagent Kit features:• Low RNA input requirements – as little as 50 ng total RNARead more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
90241510 Reactions
90241630 Reactions
Catalog number 902415
Price (USD)
1,742.00
Each
Quantity:
10 Reactions
Price (USD)
1,742.00
Each
The GeneChip™ 3' IVT PLUS Reagent Kit features:
• Low RNA input requirements – as little as 50 ng total RNA input for a single round of amplification to leave more samples for further studies
• Expression assay kit – GeneChip 3' IVT Express Kit (3' IVT Express Kit) for continuation of high-quality of data generation without compromising the legacy data
• Protocol optimization with no initial primer annealing or cDNA purification step for a streamLined workflow
• Single-day protocol with appropriate inputs of total RNA for rapid sample processing
• Magnetic bead purification for enhanced recovery to generate sufficient target
• Built-in controls to assist in effective trouble shooting
With enhanced performance, fewer required reagents, and a simple protocol, target preparation has never been so easy.
3' IVT PLUS Kit includes:
• All reagents needed to generate and label aRNA target
• Ready to use magnetic bead purification for enhanced recovery
• Poly-A RNA controls
• Hyb controls
• Tubes organizer for quick assay set up
About Poly-A controls
Poly-A controls (lys, phe, thr, and dap) monitor the target labeling process from start to finish. They serve as sensitivity indicators of target prep and labeling efficiency. This kit contains four exogenous, pre-mixed, poly-adenylated prokaryotic controls that are spiked directly into RNA samples before target labeling. Their resultant signal intensities on Applied Biosystems GeneChip Arrays serve as indicators of the labeling reaction efficiency.
About Hyb controls
The 20X Eukaryotic hybridization controls are comprised of a mixture of biotinylated and fragmented cRNA of bioB, bioC, bioD, and cre in staggered concentrations, ready to be added directly to the hybridization cocktail. These controls facilitate monitoring of the hybridization process for troubleshooting. Control Oligo B2 is included to provide alignment signals for image analysis.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
For Use With (Application)Transcriptome Profiling
Product LineGeneChip
Quantity10 Reactions
Reagent TypeSample preparation reagents
Type3' IVT PLUS Reagent Kit
Unit SizeEach
Frequently asked questions (FAQs)
What reagent kit should I use with my array?
Please refer to the Microarray Reagent Guide for Arrays and Expression Kits to match the correct reagents your array.
Find additional tips, troubleshooting help, and resources within our Microarray Analysis Support Center.
Where can I download the control sequences for expression arrays?
Sequences of all our controls can be downloaded from the sequence file that is available on product pages on thermofisher.com.
For example, here is the link to the product page for Clariom D Assay, human: https://www.thermofisher.com/order/catalog/product/902923
Go to the "Software & Data Analysis" section and under "Support Files", click on "Sequence Files: Clariom D Array, human Probe Sequences, tabular format", which you can open in Excel.
Note: If the control sequence file is not available, please contact techsupport@thermofisher.com.
Are pseudogene databases included in the design of expression arrays?
Pseudogene databases were not included in the design of expression arrays.
What is the shelf life of the Poly-A RNA Control's first dilution during processing of expression microarrays?
The first dilution of the Poly-A RNA Control can be stored at -20 degrees C for 6 weeks (with up to 4 freeze/thaws).
How long can I store labeled cDNA when working with expression microarrays?
Labeled material can be stored for 2 weeks at -20 degrees C.