GeneChip™ Human Genome U133 Plus 2.0 Assay, 10 reactions - FAQs
GeneChip™ Human Genome U133 Plus 2.0 Assay, 10 reactions - FAQs
View additional product information for GeneChip™ Human Genome U133 Plus 2.0 Assay - FAQs (902482, 902483)
3 product FAQs found
How is this array design principle different from the GeneChip Human Genome U133 type of arrays?
The GeneChip Exon Arrays represent a new array design philosophy for the most comprehensive and informative coverage with the exon as the basic unit of expression analysis. Such an inclusive design enables discovery of new and novel splicing events not previously observed experimentally. Some of the key differences are summarized below using the human arrays as an example; refer to the “GeneChip Exon Array Design” Technical Note and the “Design and Performance of the GeneChip Human Genome U133 Plus 2.0 and Human Genome U133A 2.0 Array” Technical Note for more details.
Find additional tips, troubleshooting help, and resources within our Microarray Analysis Support Center.
On average, how many probes are used to represent a corresponding GeneChip Human Genome U133 (HG-U133) Array probe set where there are 11 probe pairs?
The majority of the HG-U133 Plus 2.0 Array consensus sequences are well covered in the new design with approximately 75% of them containing over 8 probes on the Exon Array. The remaining 25% have less overlap with the new exon probes, and are mostly attributed to EST-based HG-U133 Array probe sets that do not contain an annotated coding region. As a result, in many such cases, the entire HG-U133 Array EST-based consensus sequence is designated as one PSR, with around 4 probes selected for that entire region.
Find additional tips, troubleshooting help, and resources within our Microarray Analysis Support Center.