GeneChip™ Human Gene 2.0 ST Array
Applied Biosystems™
GeneChip™ Human Gene 2.0 ST Array
The GeneChip™ Human Gene 2.0 ST Array provides the most accurate, sensitive, and comprehensive measurement of protein coding and long자세히 알아보기
Have Questions?
보기 방식 변경buttonViewtableView
카탈로그 번호어레이 수
9024992 arrays
9021126 arrays
90211330 arrays
카탈로그 번호 902499
제품 가격(KRW)
681,000
Each
어레이 수:
2 arrays
제품 가격(KRW)
681,000
Each
The GeneChip™ Human Gene 2.0 ST Array provides the most accurate, sensitive, and comprehensive measurement of protein coding and long intergenic non-coding RNA transcripts.
Comprehensive design
Keeping pace with the research community's understanding of the transcriptome, we have designed whole-transcript arrays that include probes to measure both messenger (mRNA) and long intergenic non-coding RNA transcripts (lincRNA). These whole-transcript array designs provide a complete expression profile of mRNA as well as the intermediary lincRNA transcripts that impact the mRNA expression profile.
Research over the past 20 years has predominantly focused on protein coding messenger RNA transcripts and their role in cellular processes, such as disease and development. Recently researchers have identified more that 10,000 transcripts (>200 bases) with little or no protein coding potential. Only a small fraction of these non-coding RNAs has functional annotations to date. However, there is ample evidence that differential expression of lincRNAs plays an important role in the genesis and progression of disease and that aberrant expression of these molecules have also been linked to cancer. Recent advancements in transcriptome profiling provided evidence of the association of lincRNAs in a diverse range of cellular functions:
• Regulation of mRNA transcription
• Regulation of mRNA post-transcriptional modifications
• Occlusion/recruitment of transcription factor binding
• Activation and transportation of transcription factors
• Interaction with accessory proteins
• Guide protein complexes to locations in the genome
Key benefits
• Comprehensive coverage provides the best opportunity to discover interesting biology
• >30,000 coding transcripts
• >11,000 long intergenic non-coding transcripts
• Reproducible: Intra-lot correlation coefficient =0.99
Content profile
Since the design of the Human Gene 1.0 ST Array, there have been a massive number of new lincRNA that have been identified by the research community. In order to provide the research community with a tool that can measure the differential expression of this exciting class of RNA transcripts, we have designed the Human Gene 2.0 ST Array. To supplement the lincRNA data contained in RefSeq, we used sequences and transcripts from lncRNA db www.lncrnadb.com and Broad Institute, Human Body Map lincRNAs and TUCP (transcripts of uncertain coding potential catalog) lincRNAs
For Research Use Only. Not for use in diagnostic procedures.
사양
포함Probe
제품라인GeneChip
수량2 arrays
유형Human Gene 2.0 ST Array
어레이Transcriptome Profiling
형식Array Cartridge
어레이 수2 arrays
Human
Unit SizeEach
자주 묻는 질문(FAQ)
What reagent kit should I use with my array?
Please refer to the Microarray Reagent Guide for Arrays and Expression Kits to match the correct reagents your array.
Find additional tips, troubleshooting help, and resources within our Microarray Analysis Support Center.
Are pseudogene databases included in the design of expression arrays?
Pseudogene databases were not included in the design of expression arrays.
How long can I store labeled cDNA when working with expression microarrays?
Labeled material can be stored for 2 weeks at -20 degrees C.