GeneChip™ Human Transcriptome Pico Assay 2.0, 12 reactions - FAQs
GeneChip™ Human Transcriptome Pico Assay 2.0, 12 reactions - FAQs
View additional product information for GeneChip™ Human Transcriptome Pico Assay 2.0 - FAQs (902661, 902662)
2 product FAQs found
Is there a recommended RIN (RNA integrity number), based on Bioanalyzer, for RNA quality for IVT and WT assays?
For IVT assays, the RIN should be 6 or higher. For WT assays, there are no recommendations based on assay priming.
Find additional tips, troubleshooting help, and resources within our Microarray Analysis Support Center.