Clariom™ S Pico Assay HT, rat, 96 reactions - FAQs
Clariom™ S Pico Assay HT, rat, 96 reactions - FAQs
View additional product information for Clariom™ S Pico Assay HT, rat - FAQs (902968, 902967)
1 product FAQs found