2-Propanol, ≥99.0%, USP, Multi-Compendial, GMP, J.T.Baker™
Avantor™

2-Propanol, ≥99.0%, USP, Multi-Compendial, GMP, J.T.Baker™

This product is GMP Manufactured and meets JP, BP, USP, and EP Requirements.
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
4 L
20 L
345 lb.
Catalog number 9037-01
Price (KRW)
-
Quantity:
500 mL
This product is GMP Manufactured and meets JP, BP, USP, and EP Requirements.
Specifications
Boiling Point82°C (1013 hPa)
CAS67-63-0
Chemical Name or Material2-Propanol
ColorColorless
Melting Point-89°C
Molecular Formula(CH3)2CHOH
PackagingAmber glass bottle, narrow mouth
SynonymIsopropyl alcohol, Isopropyl rubbing alcohol, sec-Propanol, sec-Propyl alcohol, 2-Hydroxy propane
GradeMulti-Compendial/USP
Molecular Weight (g/mol)60.1
Percent Purity≥99.0%
Quantity500 mL
Unit SizeEach