2-Propanol, ≥99.0%, USP, Multi-Compendial, GMP, J.T.Baker™
Avantor™

2-Propanol, ≥99.0%, USP, Multi-Compendial, GMP, J.T.Baker™

This product is GMP Manufactured and meets JP, BP, USP, and EP Requirements.
Have Questions?
보기 방식 변경buttonViewtableView
수량:
500 mL
4 L
20 L
345 lb.
카탈로그 번호 9037-20
제품 가격(KRW)
-
수량:
345 lb.
This product is GMP Manufactured and meets JP, BP, USP, and EP Requirements.
사양
끓는점82°C (1013 hPa)
CAS67-63-0
CAS67-63-0
화학물질 이름 또는 재질2-Propanol
색상Colorless
용융점-89°C
분자식(CH3)2CHOH
동의어Isopropyl alcohol, Isopropyl Rubbing Alcohol, sec-Propanol, sec-Propyl Alcohol, 2-Hydroxy propane
등급Multi-Compendial/USP
분자량(g/mol)60.1
백분율 순도≥99.0%
수량345 lb.
Unit SizeEach