CD14 APC-eFluor 780, CE-IVD, eBioscience™ - FAQs
CD14 APC-eFluor 780, CE-IVD, eBioscience™ - FAQs
View additional product information for CD14 APC-eFluor 780, CE-IVD, eBioscience™ - FAQs (9047-0149-120)
1 product FAQs found
Can the eFluor Organic Dyes be frozen?
As with other organic fluorochromes, we do not recommend the eFluor Organic Dyes be frozen.