GeneChip™ PrimeView™ Global Gene Expression Profile Assay - FAQs
GeneChip™ PrimeView™ Global Gene Expression Profile Assay - FAQs
View additional product information for GeneChip™ PrimeView™ Global Gene Expression Profile Assay - FAQs (905400)
7 product FAQs found
Which kit do I need to purchase to run the PluriTest Assay?
Customers should purchase the GeneChip PrimeView Global Gene Expression Profile Assay (Cat. No. 905406 or 905407). 
Find additional tips, troubleshooting help, and resources within our Microarray Analysis Support Center.
What additional reagents do I need to run the GeneChip PrimeView Global Gene Expression Profile Assay, Cat. No. 905406 or 905407?
The GeneChip PrimeView Global Gene Expression Profile Assay (Cat. No. 905406 or 905407) does not include the hybridization, wash, and stain reagents. Hence, the GeneChip Hybridization, Wash, and Stain Kit (Cat. No. 900720) will have to be purchased separately. Alternatively, the GeneChip PrimeView Global Gene Expression Profile Assay (Cat. No. 905400) can be used; it is a complete kit that includes the GeneChip Hybridization, Wash, and Stain kit. 
Find additional tips, troubleshooting help, and resources within our Microarray Analysis Support Center.
What is the relationship between the inventors of PluriTest and Thermo Fisher Scientific?
Thermo Fisher Scientific has an exclusive global license from Drs. Jeanne Loring and Franz Joseph Muller under the Intellectual Property rights related to PluriTest.
I am having problems accessing PluriTest or I am unable to upload data for analysis. What do you suggest?
Check the web browser you are using. PluriTest is designed for use with most recent versions of the following browsers: Chrome on Windows or Mac OS X, Microsoft Edge on Windows 10, and Firefox on Windows or Mac OS X.
Do I need to register to use PluriTest online analysis?
Yes, please visit https://www.pluritest2.org/ to create a user account by clicking on the Register icon located on the upper right corner of the homepage.
Which PrimeView Gene Expression Assays are compatible with PluriTest?
GeneChip PrimeView Global Gene Expression Profile Assay (Cat. No. 905400) and PrimeView 16 Global Gene Expression Profile Assay (Cat. No. 905402) are compatible with PluriTest. GeneChip PrimeView Global Gene Expression Profile Assay (Cat. No. 905400) is run on the GeneChip Scanner 3000 instrument. PrimeView 16 Global Gene Expression Profile Assay (Cat. No. 905402) is run on the GeneTitan family of instruments.
What are PrimeView Gene Expression Assays?
The PrimeView Global Gene Expression Profile Assays enable expression with an emphasis on established, well-annotated content. More than 36,000 transcripts and variants are measured per sample. Sequences used in the design of the assay were selected from the UniGene database 219, RefSeq version 36, and full-length human mRNAs from GenBank database.