PrimeView™ 16 Global Gene Expression Profile Assay
Applied Biosystems™

PrimeView™ 16 Global Gene Expression Profile Assay

The PrimeView 16 Global Gene Expression Profile Assay enables expression profiling of 16 samples at a time with an emphasis on established, well-annotated content.
Have Questions?
Catalog NumberQuantity
90540216 assays
Catalog number 905402
Price (USD)
-
Quantity:
16 assays

The PrimeView 16 Global Gene Expression Profile Assay enables expression profiling of 16 samples at a time with an emphasis on established, well-annotated content. More than 36,000 transcripts and variants are measured per sample. Sequences used in the design of the array were selected from the UniGene database 219, RefSeq version 36, and full-length human mRNAs from GenBank™.

Note: The PluriTest™ Assay now accepts PrimeView array data from GeneTitan instruments.

Features include:

  • Cost-effective global confirmation of pluripotency marker expression
  • Simple-to-use online analysis tool
  • Published method with over 15,000 samples analyzed
  • For use with GeneTitan family of instruments

About the PluriTest Assay
The PluriTest Assay is a bioinformatics assay that compares the transcriptional profile of a sample to an extensive reference set of >450 cell/tissue types, including 223 hESC lines, 41 iPSC lines, somatic cells, and tissues. The online analysis tool confirms pluripotency marker expression via two separate scores: Pluripotency and Novelty. The Pluripotency Score informs the user on how strongly a model-based pluripotency signature is expressed in the samples analyzed. The Novelty Score indicates the general model fit for a given sample.

Specifications
ArrayTranscriptome Profiling
FormatAssay
Product LineGeneChip, PrimeView
Product TypeGene Expression Profile Assay
Quantity16 assays
Shipping ConditionApproved for shipment at Room Temperature or on Wet or Dry Ice
SpeciesHuman
For Use With (Application)Stem Cell Analysis
Unit SizeEach
Contents & Storage

• Arrays (store at 2°C to 8°C)
• Purification beads, Module 2 (2°C to 8°C)
• Module 1 (-5°C to -30°C)
• PolyA kit (-5°C to -30°C)
• Hybridization kit (-5°C to -30°C)
• Fragmention buffer, Module 2 (room temperature)

Frequently asked questions (FAQs)

What is the relationship between the inventors of PluriTest and Thermo Fisher Scientific?

Thermo Fisher Scientific has an exclusive global license from Drs. Jeanne Loring and Franz Joseph Muller under the Intellectual Property rights related to PluriTest.

I am having problems accessing PluriTest or I am unable to upload data for analysis. What do you suggest?

Check the web browser you are using. PluriTest is designed for use with most recent versions of the following browsers: Chrome on Windows or Mac OS X, Microsoft Edge on Windows 10, and Firefox on Windows or Mac OS X.

Do I need to register to use PluriTest online analysis?

Yes, please visit https://www.pluritest2.org/ to create a user account by clicking on the Register icon located on the upper right corner of the homepage.

Which PrimeView Gene Expression Assays are compatible with PluriTest?

GeneChip PrimeView Global Gene Expression Profile Assay (Cat. No. 905400) and PrimeView 16 Global Gene Expression Profile Assay (Cat. No. 905402) are compatible with PluriTest. GeneChip PrimeView Global Gene Expression Profile Assay (Cat. No. 905400) is run on the GeneChip Scanner 3000 instrument. PrimeView 16 Global Gene Expression Profile Assay (Cat. No. 905402) is run on the GeneTitan family of instruments.

What are PrimeView Gene Expression Assays?

The PrimeView Global Gene Expression Profile Assays enable expression with an emphasis on established, well-annotated content. More than 36,000 transcripts and variants are measured per sample. Sequences used in the design of the assay were selected from the UniGene database 219, RefSeq version 36, and full-length human mRNAs from GenBank database.