Orion™ pH Electrode Storage Solutions
Thermo Scientific™

Orion™ pH Electrode Storage Solutions

Restore proper electrode performance and prolong electrode life using these pH electrode storage solutions.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberFor Use With (Equipment)Quantity
910001pH Electrode475 mL
810001TS
also known as 810001
ROSS pH Electrode475 mL
910060pH Electrode60 mL
9100CBpH Electrode19 L
Catalog number 910001
Price (MXN)
-
For Use With (Equipment):
pH Electrode
Quantity:
475 mL

Restore proper electrode performance and prolong electrode life using Thermo Scientific™ Orion™ pH Electrode Storage Solutions in ready to use bottles.

Storage Solutions:

  • Storage solution ensures that the pH electrode works properly after prolonged storage, overnight storage and between samples

Warranty: Out of box

Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
DescriptionpH electrode storage solution, 475 mL bottle
Filling Solution TypeStorage
For Use With (Application)Ensures that the pH electrode works properly after prolonged storage, overnight storage and between samples
PackagingBottle
Quantity475 mL
For Use With (Equipment)pH Electrode
TypepH Electrode Storage Solution
Unit SizeEach