Accessories for Orion™ pH Electrodes
Thermo Scientific™
Accessories for Orion™ pH Electrodes
Store and protect your pH electrodes with these accessories.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberFor Use With (Equipment)Type
910005Sure-Flow Electrode, Standard Glass ElectrodeBulb Guard
910004-WA8 mm Electrode, GlassElectrode Storage Bottle
910006-WA6 mm Electrode, GlassElectrode Storage Bottle
910003-WA12 mm Electrodes, GlassElectrode Storage Bottle
Catalog number 910005
Price (USD)
53.37
Online exclusive
59.30
Save 5.93
Pack of 5
For Use With (Equipment):
Sure-Flow Electrode, Standard Glass Electrode
Type:
Bulb Guard
Price (USD)
53.37
Online exclusive
59.30
Save 5.93
Pack of 5
Store and protect your pH electrodes with the Thermo Scientific™ Accessories for Orion™ pH Electrodes. Storage bottles protect and condition electrodes when not in use. Bulb guard protects the pH-sensing bulb of glass pH electrodes.
Electrode Storage Bottles
  • Can be used with any 6, 8 or 12mm pH electrodes
Electrode Bulb Guards
  • Designed for glass body pH electrodes
  • Use with standard or Thermo Scientific™ Orion™ Sure-Flow™ pH electrodes — one end of the bulb guard fits over standard electrodes while the other end fits over Sure-Flow electrodes without interfering with the Sure-Flow flushing action
Specifications
DescriptionBulb guard for glass pH Electrodes
For Use With (Equipment)Sure-Flow Electrode, Standard Glass Electrode
TypeBulb Guard
Unit SizePack of 5