Orion™ Electrode Extension Cables
Thermo Scientific™
Orion™ Electrode Extension Cables
Extend the length of an electrode cable by 15' (4.57m) when using the Thermo Scientific™ Orion™ Electrode Extension Cables.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberConnector Types
910027BNC
910026-WAPin Tip
9100328 Pin MiniDIN
Catalog number 910027
Price (USD)
240.30
Online Exclusive
267.00
Save 26.70 (10%)
Each
Connector Types:
BNC
Price (USD)
240.30
Online Exclusive
267.00
Save 26.70 (10%)
Each
Extend the length of an electrode cable by 15' (4.57m) when using the Thermo Scientific™ Orion™ Electrode Extension Cables. Can be used with Orion ATC probes, any reference electrode with 2mm pin tip connector or any electrode with a BNC connector.
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
Connector TypesBNC
Description15 ft. Extension Cable, BNC connector
Length (English) Cable15 ft.
Length (Metric) Cable4.57 m
For Use With (Equipment)Meters and Probes with BNC Connectors
TypeElectrode Extension Cable
Unit SizeEach