Orion™ Electrode Extension Cables
Actual product may vary
Orion™ Electrode Extension Cables
Orion™ Electrode Extension Cables
Thermo Scientific™
Orion™ Electrode Extension Cables
Extend the length of an electrode cable by 15' (4.57m) when using the Thermo Scientific™ Orion™ Electrode Extension Cables.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberConnector Types
9100328 Pin MiniDIN
910026-WAPin Tip
910027BNC
Catalog number 910032
Price (USD)
218.70
Online Exclusive
243.00
Save 24.30 (10%)
Each
Connector Types:
8 Pin MiniDIN
Price (USD)
218.70
Online Exclusive
243.00
Save 24.30 (10%)
Each
Extend the length of an electrode cable by 15' (4.57m) when using the Thermo Scientific™ Orion™ Electrode Extension Cables. Can be used with Orion ATC probes, any reference electrode with 2mm pin tip connector or any electrode with a BNC connector.
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
Connector Types8 Pin MiniDIN
Description15 ft. Extension Cable, 8 Pin MiniDIN for ATC Probes Used on Star Meters
Length (English) Cable15 ft.
Length (Metric) Cable4.57 m
For Use With (Equipment)ATC Probes Used on Star Meters
TypeElectrode Extension Cable
Unit SizeEach