Orion™ Double Junction Glass Bodied Combination pH Electrode
Orion™ Double Junction Glass Bodied Combination pH Electrode
Orion™ Double Junction Glass Bodied Combination pH Electrode
Orion™ Double Junction Glass Bodied Combination pH Electrode
Orion™ Double Junction Glass Bodied Combination pH Electrode
Thermo Scientific™

Orion™ Double Junction Glass Bodied Combination pH Electrode

Capture accurate measurements of a variety of samples types, including solvents, with Orion glass body pH electrodes.
Have Questions?
Catalog NumberConnector Types
9102DJWPBNC (Waterproof)
Catalog number 9102DJWP
Price (MXN)
-
Connector Types:
BNC (Waterproof)
Thermo Scientific™ Orion™ Silver Chloride Double Junction Refillable Electrodes provide reliable and accurate measurements of routine samples.They are available in chemically resistant glass bodies and best suited for general purpose samples, treated effluent and distilled water.
 

Orion™ Double Junction Electrodes have an isolated Ag/AgCl reference system which prevents silver from coming in contact with the sample. Great for measuring TRIS buffer, sulfide and protein samples.

Solutions required

  • Electrode fill solution
  • 910008 3M KCl filling solution, 5 x 60 mL bottles

Electrode storage solution

  • 910001 pH electrode storage solution, 475 mL bottle

pH buffers

  • 910168 pH 1.68 buffer, NIST traceable, 475 mL
  • 910104 pH 4.01 buffer, color coded red, NIST traceable, 475 mL
  • 910105 pH 5.00 buffer, color coded orange, NIST traceable, 475 mL
  • 910686 pH 6.86 buffer, DIN standard, NIST traceable, 475 mL
  • 910107 pH 7.00 buffer, color coded yellow, NIST traceable, 475 mL
  • 910918 pH 9.18 buffer, DIN standard, NIST traceable, 475 mL
  • 910110 pH 10.01 buffer, color coded blue, NIST traceable, 475 mL
  • 910112 pH 12.46 buffer, NIST traceable, 475 mL
  • 910199 Standard All-in-One pH buffer kit, includes pH 4.01, 7.00 and 10.01 buffers, 475ml each, pH
  • electrode storage solution, 475 mL, and 12 mm electrode storage bottle

pH cleaning solution kit

  • 900020 Cleaning solution A and C, 30 mL each. B and D, 60 mL each

Items included with

  • 910008 3M KCl filling solution 60 mL 1 bottle

Warranty period

  • 12 months from date of purchase
Specifications
DescriptionDouble Junction pH Electrode
Diameter (English) Body0.47 in.
Diameter (Metric) Body12 mm
Filling Solution Type3M KCl
For Use With (Equipment)Standard pH Meter
Internal ReferenceAg/AgCl Double Junction
Length (English)4.7 in.
Length (Metric)120 mm
Precision0.02
Storage RequirementsStore in Orion pH storage solution 910001
Temperature (English) Operating32°F to 140°F
Connector TypesBNC (Waterproof)
JunctionCeramic
MaterialGlass Body
Temperature (Metric) Operating0°C to 60°C
TypeDouble Junction pH Electrode
pH Range0 to 14
Unit SizeEach