Orion™ Single Junction Thin Stem Glass Bodied Combination pH Electrode
Thermo Scientific™

Orion™ Single Junction Thin Stem Glass Bodied Combination pH Electrode

Top-performance, semi-micro pH electrodes to measure samples as small as 0.2 mL
Have Questions?
Catalog NumberConnector Types
9103BNWPBNC (Waterproof)
Catalog number 9103BNWP
Price (USD)
407.70
Online exclusive
453.00
Save 45.30
Each
Connector Types:
BNC (Waterproof)
Price (USD)
407.70
Online exclusive
453.00
Save 45.30
Each
Thermo Scientific™ Orion™ Silver Chloride Semi-Micro Electrodes provide reliable and accurate measurements of routine samples. They are available in chemically resistant glass bodies and best suited for measuring general samples in test tubes and small containers.
 

Orion™ Single Junction Electrodes provide a reliable option for routine pH measurement. These silver chloride electrodes are available in a variety of styles and provide accurate measurements of typical samples.

Solutions required

Electrode storage solution

  • 910001 pH electrode storage solution, 475 mL bottle

pH buffers

  • 910168 pH 1.68 buffer, NIST traceable, 475 mL
  • 910104 pH 4.01 buffer, color coded red, NIST traceable, 475 mL
  • 910105 pH 5.00 buffer, color coded orange, NIST traceable, 475 mL
  • 910686 pH 6.86 buffer, DIN standard, NIST traceable, 475 mL
  • 910107 pH 7.00 buffer, color coded yellow, NIST traceable, 475 mL
  • 910918 pH 9.18 buffer, DIN standard, NIST traceable, 475 mL
  • 910110 pH 10.01 buffer, color coded blue, NIST traceable, 475 mL
  • 910112 pH 12.46 buffer, NIST traceable, 475 mL
  • 910199 Standard All-in-One pH buffer kit, includes pH 4.01, 7.00 and 10.01 buffers, 475ml each, pH electrode storage solution, 475ml, and 12 mm electrode storage bottle

pH cleaning solution kit

  • 900020 Cleaning solution A and C, 30 mL each. B and D, 60 mL each

Warranty period

  • 6 months from date of purchase
Specifications
DescriptionWith water proof connector
Diameter (Metric) Cap16 mm
Filling Solution Type4M KCl with Ag/AgCl
For Use With (Equipment)Standard pH Meter, BNC connector
Internal ReferenceAg/AgCl
Precision0.02
Temperature (English) Operating32°F to 194°F
Connector TypesBNC (Waterproof)
JunctionCeramic
MaterialGlass Body
Temperature (Metric) Operating0°C to 90°C
TypeCombination Ag/AgCl pH Electrode, glass body, BNC waterprood connector
pH Range0 to 14
Unit SizeEach