Orion™ pH Buffer Individual Use Pouches
Orion™ pH Buffer Individual Use Pouches
Orion™ pH Buffer Individual Use Pouches
Orion™ pH Buffer Individual Use Pouches
Orion™ pH Buffer Individual Use Pouches
Orion™ pH Buffer Individual Use Pouches
Orion™ pH Buffer Individual Use Pouches
Thermo Scientific™

Orion™ pH Buffer Individual Use Pouches

Orion™ single-use pH calibration buffers deliver NIST-traceable accuracy for pH meters and electrodes. Designed for GxP labs, these ready-to-use, color-coded buffer pouches support compliant workflows, reduce contamination risk and ensure reliable, traceable pH calibration results.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantitypH
910410-WA10/Pk.4.01
91042525/Pk.4.01
91071010/Pk.7
91072525/Pk.7
91101010/Pk.10.01
911025-WA25/Pk.10.01
Catalog number 910410-WA
Price (USD)
44.73
Online exclusive
49.70
Save 4.97
Case of 10
Quantity:
10/Pk.
pH:
4.01
Price (USD)
44.73
Online exclusive
49.70
Save 4.97
Case of 10
Thermo Scientific™ Orion™ single-use pH calibration buffers deliver NIST-traceable accuracy for pH meters and electrodes in laboratory and GxP-regulated environments. These ready-to-use, color-coded buffer pouches support compliant calibration workflows, reduce contamination risk, and ensure consistent, reliable results. Each pouch is individually labeled for full traceability and designed for direct electrode use, improving efficiency and confidence in pH measurements.

Single-Use NIST-Traceable pH Calibration Buffers for pH Meter and Electrode Calibration

Each pouch is individually labeled with catalog number, lot code and expiration date, supporting documentation and traceability required for GMP, GLP and ISO quality systems.

pH 4, 7, and 10 buffer pouches and solutions are color-coded for quick identification during calibration.

  • pH 4 buffer pouches and solution are light red
  • pH 7 buffer pouches and solution are yellow
  • pH 10 buffer pouches and solution are blue

Redesigned pouches offer a rectangular shape to better accommodate electrodes placed directly into the pouch and 20 mL solution volume to ensure proper electrode immersion for accurate calibration.

Key Features

  • pH buffers are traceable to NIST reference standards
  • Each pH buffer pouch and solution is color-coded
  • Each pouch is labeled with catalog number, lot code and expiration date
  • Each pouch is easy to open with tear notches
  • Each pouch is single use to reduce contamination risk

Workflow and Application Benefits

Orion pH buffer individual-use pouches simplify pH calibration procedures in both laboratory and field settings.

  • Direct electrode calibration: Updated pouch design allows electrodes to be placed directly into the pouch for quick calibration steps.
  • Quick identification during calibration: Color-coded buffers help technicians rapidly identify the correct standard during multi-point calibration procedures.
  • Consistent calibration performance: Single-use pouches ensure each calibration uses fresh buffer solution, reducing variability caused by contamination or evaporation.
  • Convenient calibration anywhere: Compact pouches are ideal for field testing, environmental monitoring and routine QA/QC calibration workflows.
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
Boiling Point100°C
Certificate of AnalysisYes
Certifications/ComplianceISO 9001
Chemical Name or MaterialpH Buffer, individual use pouches
ColorPink
FormulationWater, Potassium Hydroxide, Amaranth Red No. 27, Potassium Hydrogen Phthalate
PackagingPouch
Solubility InformationSoluble
Traceability to NISTYes
For Use With (Application)pH calibration
For Use With (Equipment)pH Meter and Electrode, single-use
GradeCertified
Physical FormLiquid
Quantity10/Pk.
Solution TypeBuffer
pH4.01
Unit SizeCase of 10