Orion™ AquaPro™ Double Junction Combination pH Electrodes
Thermo Scientific™

Orion™ AquaPro™ Double Junction Combination pH Electrodes

Measure the pH of viscous liquid samples quickly, and accurately, with clog-resistant Orion™ AquaPro™ pH electrodes.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberConnector TypesMaterialTypeTip Type
9107APMDMini-DIN (Star)Epoxy BodypH/ATC Combination ElectrodeStandard
9104APWPBNC (Waterproof)Glass BodyCombination pH ElectrodeToughened Bulb
9135APWPBNC (Waterproof)Epoxy BodyCombination pH ElectrodeFlat Surface
9156APWPBNC (Waterproof)Epoxy BodyCombination pH ElectrodeStandard
Catalog number 9107APMD
Price (USD)
360.90
Online exclusive
401.00
Save 40.10
Each
Connector Types:
Mini-DIN (Star)
Material:
Epoxy Body
Type:
pH/ATC Combination Electrode
Tip Type:
Standard
Price (USD)
360.90
Online exclusive
401.00
Save 40.10
Each
Thermo Scientific™ Orion™ AquaPro™ Low Maintenance pH Electrodes provide fast response and extended life with the additional advantages of a double junction design. Their clog resistant, open junctions allow for use in difficult samples, while making post-use cleaning, simple. They are available in a variety of different designs, with the standard types being the 9104APWP & the 9156APWP. These have glass and epoxy bodies respectively and are designed for general lab use. The 9135APWP has a uniquely designed, flat sensing bulb that allows for reliable pH measurement on samples such as paper, textiles, skin, meat, cheese, or in agar gel plates. The 9107APMD contains a built-in temperature sensor that accurately measures pH and temperature with just one electrode, so combines the fast response and open junction design of the Orion™ AquaPro™ Electrode with a built-in ATC.
 

Orion™ AquaPro™ Electrodes have polymer filled, double junction reference systems that are low maintenance. They have an extended life, fast response and clog resistant open junctions and are ideal for use in Tris, sulfides, proteins and biological media. The junction must be kept wet when not in use.

Solutions required

Electrode storage solution

  • 910001 pH electrode storage solution, 475 mL bottle

pH buffers

  • 910168 pH 1.68 buffer, NIST traceable, 475 mL
  • 910104 pH 4.01 buffer, color coded red, NIST traceable, 475 mL
  • 910105 pH 5.00 buffer, color coded orange, NIST traceable, 475 mL
  • 910686 pH 6.86 buffer, DIN standard, NIST traceable, 475 mL
  • 910107 pH 7.00 buffer, color coded yellow, NIST traceable, 475 mL
  • 910918 pH 9.18 buffer, DIN standard, NIST traceable, 475 mL
  • 910110 pH 10.01 buffer, color coded blue, NIST traceable, 475 mL
  • 910112 pH 12.46 buffer, NIST traceable, 475 mL
  • 910199 Standard All-in-One pH buffer kit, includes pH 4.01, 7.00 and 10.01 buffers, 475ml each, pH electrode storage solution, 475ml, and 12 mm electrode storage bottle

pH cleaning solution kit

  • 900020 Cleaning solution A and C, 30 ml each. B and D, 60 mL each

Warranty period

  • 12 months from date of purchase
Specifications
DescriptionEpoxy Body pH/ATC
Diameter (English) Body0.47 in.
Diameter (Metric) Body12 mm
For Use With (Application)General Purpose
For Use With (Equipment)Orion Star Series Meters
Internal ReferenceAg/AgCl Double Junction
Length (English)4.72 in.
Length (English) Cable39 in.
Length (Metric)120 mm
Length (Metric) Cable1 m
Storage RequirementsAlways store in pH storage solution, Orion 910001. Either store in a beaker of storage solution or reassemble in the storage bottle the electrode was provided with.
Temperature (English) Operating32°F to 140°F
Tip TypeStandard
Connector TypesMini-DIN (Star)
JunctionOpen
MaterialEpoxy Body
Temperature (Metric) Operating0°C to 60°C
TypepH/ATC Combination Electrode
pH Range0 to 14
pH Relative Accuracy±0.02
Unit SizeEach