Thermo Scientific™
Rheodyne™ Suction Needle Adapter
 Related applications:
Industrial Chromatography
Use the Thermo Scientific™ Rheodyne™ Suction Needle Adapter for suction loading of samples into the Rheodyne 9725 and 9725i Sample Injectors for high-performance liquid chromatography (HPLC),
Error loading your content!
  Catalog number Price ({{currency}}) Availability Qty
{{product.sku}}
also known as {{product.formattedSku}} 
{{product.availability.message}} Add To Cart
{{addItemToList.successMessage}} {{addItemToList.listName}}
Back to top
Description
Use the Thermo Scientific™ Rheodyne™ Suction Needle Adapter for suction loading of samples into the Rheodyne 9725 and 9725i Sample Injectors for high-performance liquid chromatography (HPLC),
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Documents
Manuals & protocols