Quantofix™ Peroxide 25 Semi-quantitative Test Strips
Macherey-Nagel™

Quantofix™ Peroxide 25 Semi-quantitative Test Strips

Ideal to test low levels of peroxide in solutions. Macherey Nagel™ Quantofix™ Peroxide 25 Semi-quantitative Test Strips use an easy dip-and-read procedure and provide reliable results within 15 seconds.
Have Questions?
Catalog NumberColor Indicator
91319TS
also known as 91319
White to Blue
Catalog number 91319TS
also known as 91319
Price (USD)
-
Color Indicator:
White to Blue
Ideal to test low levels of peroxide in solutions. Macherey Nagel™ Quantofix™ Peroxide 25 Semi-quantitative Test Strips use an easy dip-and-read procedure and provide reliable results within 15 seconds.
  • Rapid: most tests can be carried out in 10s to 120s
  • Convenient: ready to use kits, pre-calibrated, contain all necessary equipment and reagents
  • Precise: color charts adjusted and checked using certified standard solutions
  • Shelf life: 2. 5 years, store below 30°C
Specifications
Color IndicatorWhite to Blue
For Use With (Application)Determination of Peroxide 25
FormatStrip
Graduations0, 0.5, 2, 5, 10, 25mg/L H2O2
Quantity100 Pack
Shelf Life2.5 Years
Storage Requirements30°C
Test Time15 sec.
TypeSemi-qualitative Test Strip
Unit SizeEach