Automation Tips for Beckman™ Liquid Handling Systems with AP96 Dispense Head
Thermo Scientific™
Automation Tips for Beckman™ Liquid Handling Systems with AP96 Dispense Head
Increase pipetting performance with Thermo Scientific™ Sterile Pipette Tips for Beckman™ Liquid Handling Systems through the superior tip straightness and low %CV.
Have Questions?
Alterar visualizaçãobuttonViewtableView
Número do catálogoVolume (Metric)FilteredTip StyleFit for Purpose
916-26250 μLNon-filteredStandardCertified Quality
912-0218 μLFilteredStandardSterile and PCR Ready
912-26110 μLNon-filteredStandardSterile
912-26210 μLNon-filteredStandardCertified Quality
916-02140 μLFilteredStandardSterile and PCR Ready
916-26150 μLNon-filteredStandardSterile
916-262-0550 μLNon-filteredLow RetentionCertified Quality
918-02120 μLFilteredStandardSterile and PCR Ready
918-021-0520 μLFilteredLow RetentionSterile and PCR Ready
918-26120 μLNon-filteredStandardSterile
918-261-0520 μLNon-filteredLow RetentionSterile
918-26220 μLNon-filteredStandardCertified Quality
918-262-0520 μLNon-filteredLow RetentionCertified Quality
919-021130 μLFilteredStandardSterile and PCR Ready
919-021-05130 μLFilteredLow RetentionSterile and PCR Ready
919-021G130 μLFilteredWide BoreSterile and PCR Ready
919-261250 μLNon-filteredStandardSterile
919-261-05250 μLNon-filteredLow RetentionSterile
919-261G250 μLNon-filteredWide BoreSterile
919-262250 μLNon-filteredStandardCertified Quality
919-262-05250 μLNon-filteredLow RetentionCertified Quality
919-262G250 μLNon-filteredWide BoreCertified Quality
276050 μLFilteredExtended LengthSterile
Número do catálogo 916-262
Preço (BRL)
-
Volume (Metric):
50 μL
Filtered:
Non-filtered
Tip Style:
Standard
Fit for Purpose:
Certified Quality
Increase pipetting performance with Thermo Scientific™ Sterile Pipette Tips for Beckman™ Liquid Handling Systems, which undergo a rigorous 15-point quality control program to ensure consistency. These automation tips provide accurate and precise performance due to superior tip straightness and a low coefficient of variation. The pipette tips are validated on the actual workstations for which they are created.
Features:
  • Available with Thermo Scientific™ ART™ filter technology, low retention and extended length
  • DNase, RNase, DNA, ATP, PCR Inhibitor, and Endotoxin Free
Especificações
Certifications/ComplianceISO 9001 and ISO 13485
Clean ClaimsFree of RNase, DNase, DNA, ATP, and endotoxin
Packaging96 tips/rack, 10 racks/pack, 5 packs/case
Quantity4800 tips
AutoclavableAutoclavable
ColorClear
FilteredNon-filtered
Fit for PurposeCertified Quality
For Use With (Equipment)Beckman Coulter Biomek FX, Beckman Coulter Biomek NX, Beckman Coulter Biomek 3000, Beckman Coulter Multimek AP96
FormatRack
Product LineBeckman
SterilityNon-sterile
Tip StyleStandard
Volume (Metric)50 μL
Unit SizeCase of 4800