Electrodo especializado de punta de lanza Orion™ modelo 91-63
Thermo Scientific™

Electrodo especializado de punta de lanza Orion™ modelo 91-63

El electrodo especializado de punta de lanza Thermo Scientific™ Orion modelo 91-63 es un electrodo combinado de pH para perforar muestras blandas.
Have Questions?
Número de catálogoTipo
9163SCElectrodo de punta de lanza especial
Número de catálogo 9163SC
Precio (MXN)
-
Tipo:
Electrodo de punta de lanza especial

Electrodo combinado de pH para perforar muestras blandas.

Orion™ Single Junction Electrodes provide a reliable option for routine pH measurement. These silver chloride electrodes are available in a variety of styles and provide accurate measurements of typical samples.
 
Solutions required 
Electrode fill solution
900011     4M KCl with Ag/AgCl filling solution, 5 x 60 mL bottles
 
Electrode storage solution
910001      pH electrode storage solution, 475 mL bottle
 
pH buffers
910168     pH 1.68 buffer, NIST traceable, 475 mL
910104     pH 4.01 buffer, color coded red, NIST traceable, 475 mL
910105     pH 5.00 buffer, color coded orange, NIST traceable, 475 mL
910686     pH 6.86 buffer, DIN standard, NIST traceable, 475 mL
910107     pH 7.00 buffer, color coded yellow, NIST traceable, 475 mL 
910918     pH 9.18 buffer, DIN standard, NIST traceable, 475 mL
910110     pH 10.01 buffer, color coded blue, NIST traceable, 475 mL
910112     pH 12.46 buffer, NIST traceable, 475 mL
910199     Standard All-in-One pH buffer kit, includes pH 4.01, 7.00 and 10.01 buffers, 475ml each, pH
                 electrode storage solution, 475ml, and 12 mm electrode storage bottle
 
pH cleaning solution kit
900020     Cleaning solution A and C, 30 ml each. B and D, 60 ml each
 
Items included with
900011     4M KCl with Ag/AgCl filling solution 60mL 1 bottle
 
Warranty period
12 months from date of purchase
Especificaciones
DescripciónElectrodo de valoración especializado
TipoElectrodo de punta de lanza especial
Unit SizeEach