Automation Tips for Beckman™ Liquid Handling Systems with MP20 and MP200 Dispense Heads
Thermo Scientific™
Automation Tips for Beckman™ Liquid Handling Systems with MP20 and MP200 Dispense Heads
Increase pipetting performance with Thermo Scientific™ Automation Tips for Beckman™ Liquid Handling Systems, which undergo a rigorous 15-point quality control program to ensure consistency.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberVolume (Metric)FilteredSterilityFit for PurposeUnit Size
917-261PK
also known as 917-261
200 μLNon-filteredSterileSterilePack of 960
915-02120 μLFilteredSterileSterile and PCR ReadyCase of 4800
915-021PK
also known as 915-021
20 μLFilteredSterileSterile and PCR ReadyPack of 960
915-26120 μLNon-filteredSterileSterileCase of 4800
915-261PK
also known as 915-261
20 μLNon-filteredSterileSterilePack of 960
915-26220 μLNon-filteredNon-sterileCertified QualityCase of 4800
915-262PK
also known as 915-262
20 μLNon-filteredNon-sterileCertified QualityPack of 960
917-021130 μLFilteredSterileSterile and PCR ReadyCase of 4800
917-021PK
also known as 917-021
130 μLFilteredSterileSterile and PCR ReadyPack of 960
917-261200 μLNon-filteredSterileSterileCase of 4800
917-262200 μLNon-filteredNon-sterileCertified QualityCase of 4800
917-262PK
also known as 917-262
200 μLNon-filteredNon-sterileCertified QualityPack of 960
Catalog number 917-261PK
also known as 917-261
Price (KRW)
155,000
Pack of 960
Volume (Metric):
200 μL
Filtered:
Non-filtered
Sterility:
Sterile
Fit for Purpose:
Sterile
Unit Size:
Pack of 960
Price (KRW)
155,000
Pack of 960
Increase pipetting performance with Thermo Scientific™ Sterile Pipette Tips for Beckman™ Liquid Handling Systems, which undergo a rigorous 15-point quality control program to ensure consistency. These automation tips provide accurate and precise performance due to superior tip straightness and a low coefficient of variation. The pipette tips are validated on the actual workstations for which they are created.
Features:
  • Available with Thermo Scientific™ ART™ filter technology, low retention and extended length
  • Lot-tested during production on actual workstation for which they are created
  • Extended length tips compatible with 96-, 384-, and 1536-deep well plates
  • Low percentage coefficient of variation (%CV)
Specifications
ColorClear
FilteredNon-filtered
Fit for PurposeSterile
For Use With (Equipment)Beckman Coulter Liquid Handling Systems
FormatRack
Product LineBeckman
SterilitySterile
Tip StyleExtended Length
Volume (Metric)200 μL
Unit SizePack of 960