Orion™ Single Junction Sure-Flow™ Glass Bodied Combination pH Electrode
Orion™ Single Junction Sure-Flow™ Glass Bodied Combination pH Electrode
Orion™ Single Junction Sure-Flow™ Glass Bodied Combination pH Electrode
Orion™ Single Junction Sure-Flow™ Glass Bodied Combination pH Electrode
Orion™ Single Junction Sure-Flow™ Glass Bodied Combination pH Electrode
Orion™ Single Junction Sure-Flow™ Glass Bodied Combination pH Electrode
Thermo Scientific™

Orion™ Single Junction Sure-Flow™ Glass Bodied Combination pH Electrode

Attain stable, drift-free measurements of challenging samples using a sure-flow electrode with its unique free-flowing liquid-to-liquid junction.
Have Questions?
Catalog NumberJunction
9172BNWPSure-Flow
Catalog number 9172BNWP
Price (USD)
382.50
Online exclusive
425.00
Save 42.50
Each
Junction:
Sure-Flow
Price (USD)
382.50
Online exclusive
425.00
Save 42.50
Each
Thermo Scientific™ Orion™ Sure-Flow electrodes are built with a unique free-flowing liquid-to-liquid junction that simplifies measurements of dirty or viscous samples by helping to prevent clogging, making them easy to clean. They are available in chemically resistant glass bodies and best suited for use in strong acids/bases, pH > 12 or pH < 2, food & cosmetic emulsions, pure water and slurries.
 

Orion™ Single Junction Electrodes provide a reliable option for routine pH measurement. These silver chloride electrodes are available in a variety of styles and provide accurate measurements of typical samples.

Solutions required

  • Electrode fill solution
  • 900011 4M KCl with Ag/AgCl filling solution, 5 x 60 mL bottles

Electrode storage solution

  • 910001 pH electrode storage solution, 475 mL bottle

pH buffers

  • 910168 pH 1.68 buffer, NIST traceable, 475 mL
  • 910104 pH 4.01 buffer, color coded red, NIST traceable, 475 mL
  • 910105 pH 5.00 buffer, color coded orange, NIST traceable, 475 mL
  • 910686 pH 6.86 buffer, DIN standard, NIST traceable, 475 mL
  • 910107 pH 7.00 buffer, color coded yellow, NIST traceable, 475 mL
  • 910918 pH 9.18 buffer, DIN standard, NIST traceable, 475 mL
  • 910110 pH 10.01 buffer, color coded blue, NIST traceable, 475 mL
  • 910112 pH 12.46 buffer, NIST traceable, 475 mL
  • 910199 Standard All-in-One pH buffer kit, includes pH 4.01, 7.00 and 10.01 buffers, 475ml each, pH
  • electrode storage solution, 475ml, and 12 mm electrode storage bottle

pH cleaning solution kit

  • 900020 Cleaning solution A and C, 30 m: each. B and D, 60 ml each

Items included with

  • 900011 4M KCl with Ag/AgCl filling solution 60 mL 1 bottle

Warranty period

  • 12 months from date of purchase
Specifications
DescriptionModel 9172BNWP - Sure-Flow Junction
For Use With (Equipment)Standard pH Meter
Connector TypesBNC (Waterproof)
JunctionSure-Flow
MaterialGlass Body
Temperature (Metric) Operating0°C to 100°C
TypepH Electrode
pH Range0 to 14
Unit SizeEach