Sterile Pipette Tips for Beckman™ Liquid Handling Systems
Thermo Scientific™
Sterile Pipette Tips for Beckman™ Liquid Handling Systems
Increase pipetting performance with these sterile pipette tips, which provide accurate and precise performance due to superior tip straightness and a low coefficient of variation.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberVolume (Metric)PackagingFit for PurposeUnit Size
918-01120 μL50 x 96 tipsSterile and PCR ReadyCase of 4800
918-011PK
also known as 918-011
20 μL50 x 96 tipsSterile and PCR ReadyPack of 960
918-25120 μL10 x 96 tipsSterileCase of 4800
918-251PK
also known as 918-251
20 μL10 x 96 tipsSterilePack of 960
919-011130 μL10 x 96 tipsSterile and PCR ReadyCase of 4800
919-011PK
also known as 919-011
130 μL10 x 96 tipsSterile and PCR ReadyPack of 960
919-251250 μL50 x 96 tipsSterileCase of 4800
919-251PK
also known as 919-251
250 μL50 x 96 tipsSterilePack of 960
938-26150 μL10 x 384 tipsSterileEach
Catalog number 918-011
Price (USD)
-
Volume (Metric):
20 μL
Packaging:
50 x 96 tips
Fit for Purpose:
Sterile and PCR Ready
Unit Size:
Case of 4800
Increase pipetting performance with Thermo ScientificTM Sterile Pipette Tips for BeckmanTM Liquid Handling Systems, which undergo a rigorous 15-point quality control program to ensure consistency. These automation tips provide accurate and precise performance due to superior tip straightness and a low coefficient of variation. The pipette tips are validated on the actual workstations for which they are created.
Features:
  • Available with Thermo Scientific™ ART™ filter technology, low retention and extended length
  • Lot-tested during production on actual workstation for which they are created
  • Extended length tips compatible with 96-, 384-, and 1536-DeepWell™ pates
  • Low percentage coefficient of variation (%CV)
Warranty: A 5-year shelf life from the date of manufacture is recommended.
Specifications
Packaging50 x 96 tips
ColorBlack
FilteredFiltered
Fit for PurposeSterile and PCR Ready
For Use With (Equipment)Beckman Coulter Liquid Handling Systems
FormatRack
Product LineBeckman, Beckman
SterilitySterile
Tip StyleConductive
Volume (Metric)20 μL
Unit SizeCase of 4800