Sterile Pipette Tips for Beckman™ Liquid Handling Systems
Actual product may vary
Sterile Pipette Tips for Beckman™ Liquid Handling Systems
Sterile Pipette Tips for Beckman™ Liquid Handling Systems
Sterile Pipette Tips for Beckman™ Liquid Handling Systems
Sterile Pipette Tips for Beckman™ Liquid Handling Systems
Sterile Pipette Tips for Beckman™ Liquid Handling Systems
Sterile Pipette Tips for Beckman™ Liquid Handling Systems
Sterile Pipette Tips for Beckman™ Liquid Handling Systems
Sterile Pipette Tips for Beckman™ Liquid Handling Systems
Sterile Pipette Tips for Beckman™ Liquid Handling Systems
Sterile Pipette Tips for Beckman™ Liquid Handling Systems
Sterile Pipette Tips for Beckman™ Liquid Handling Systems
Sterile Pipette Tips for Beckman™ Liquid Handling Systems
Sterile Pipette Tips for Beckman™ Liquid Handling Systems
Thermo Scientific™
Sterile Pipette Tips for Beckman™ Liquid Handling Systems
Increase pipetting performance with these sterile pipette tips, which provide accurate and precise performance due to superior tip straightness and a low coefficient of variation.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberVolume (Metric)PackagingFit for PurposeUnit Size
919-251PK
also known as 919-251
250 μL50 x 96 tipsSterilePack of 960
918-01120 μL50 x 96 tipsSterile and PCR ReadyCase of 4800
918-011PK
also known as 918-011
20 μL50 x 96 tipsSterile and PCR ReadyPack of 960
918-25120 μL10 x 96 tipsSterileCase of 4800
918-251PK
also known as 918-251
20 μL10 x 96 tipsSterilePack of 960
919-011130 μL10 x 96 tipsSterile and PCR ReadyCase of 4800
919-011PK
also known as 919-011
130 μL10 x 96 tipsSterile and PCR ReadyPack of 960
919-251250 μL50 x 96 tipsSterileCase of 4800
938-26150 μL10 x 384 tipsSterileEach
Catalog number 919-251PK
also known as 919-251
Price (USD)
-
Volume (Metric):
250 μL
Packaging:
50 x 96 tips
Fit for Purpose:
Sterile
Unit Size:
Pack of 960
Increase pipetting performance with Thermo ScientificTM Sterile Pipette Tips for BeckmanTM Liquid Handling Systems, which undergo a rigorous 15-point quality control program to ensure consistency. These automation tips provide accurate and precise performance due to superior tip straightness and a low coefficient of variation. The pipette tips are validated on the actual workstations for which they are created.
Features:
  • Available with Thermo Scientific™ ART™ filter technology, low retention and extended length
  • Lot-tested during production on actual workstation for which they are created
  • Extended length tips compatible with 96-, 384-, and 1536-DeepWell™ pates
  • Low percentage coefficient of variation (%CV)
Warranty: A 5-year shelf life from the date of manufacture is recommended.
Specifications
Packaging50 x 96 tips
FilteredNon-filtered
Fit for PurposeSterile
For Use With (Equipment)Beckman Coulter Liquid Handling Systems
FormatRack
Product LineBeckman
SterilitySterile
Tip StyleConductive
Volume (Metric)250 μL
Unit SizePack of 960