Thermo Scientific™
HiPur™ Denatured Ethanol
Related applications:
Anatomical Pathology
Choose the option that best fits your work, Thermo Scientific™ HiPur™ Denatured Ethanol is available in one, five and 55 gallon sizes.
Back to top
  • Maximum of 0.1% water
  • Maximum residue content of 0.003%
  • Anhydrous formula is denatured with Methanol, MIBK, Ethyl Acetate and Heptane