Racks for Forma™ 900, 7000 Series and TSE/TSD Series Freezer
Racks for Forma™ 900, 7000 Series and TSE/TSD Series Freezer
Racks for Forma™ 900, 7000 Series and TSE/TSD Series Freezer
Racks for Forma™ 900, 7000 Series and TSE/TSD Series Freezer
Thermo Scientific™

Racks for Forma™ 900, 7000 Series and TSE/TSD Series Freezer

Forma™ 900, 7000 Series and TSE/TSD Series Freezer용 Thermo Scientific™ Rack으로 초저온 냉동고의 보관 공간을 극대화하십시오.
Have Questions?
보기 방식 변경buttonViewtableView
카탈로그 번호보유위치제품 유형용도(장비)
920022Boxes (2 in.)22 Boxes/RackSide Access Freezer Rack (Adjustable)17.3, 23 cu. ft. Upright Freezers
830030General Freezer Items5Specialty Rack - 8-Compartment Container13 cu. ft. Upright Freezers
920015Boxes (2 in.)15 Boxes/RackSide Access Freezer Rack (Adjustable)13 cu. ft. Upright Freezers
920018Boxes (2 in.)20 Boxes/RackSide Access Freezer Rack (Adjustable)17.3, 23 cu. ft. Upright Freezers
920094Boxes (2 in.)5Box Racks - Sliding Drawer13 cu. ft. Upright Freezers
920096Boxes (2 in.)5Box Racks - Sliding Drawer17.3, 23 cu. ft. Upright Freezers
920097Boxes (3 in.)12 Boxes/RackBox Racks - Sliding Drawer17.3, 23 cu. ft. Upright Freezers
398329Boxes (2 in.)20 Boxes/RackBox Racks - Sliding DrawerThermo Scientific TDE and TSX Series
1950518Boxes (2 in.)20 Boxes/RackSide Access Freezer Rack (Adjustable)Thermo Scientific TDE and TSX Series (5 shelves)
1950519Boxes (3 in.)15 Boxes/RackSide Access Freezer Rack (Adjustable)Thermo Scientific TDE and TSX Series (5 shelves)
1950522Microplates or Deepwell Microplates77 Plates/RackSide Access Freezer Rack (Microplate/Deepwell)Thermo Scientific TDE and TSX Series (5 shelves)
398328Boxes (3 in.)15 Boxes/RackBox Racks - Sliding DrawerThermo Scientific TDE and TSX Series (5 shelves)
820030Microplates30Freezer RackForma Upright ULT Freezers
카탈로그 번호 920022
제품 가격(KRW)
-
보유:
Boxes (2 in.)
위치:
22 Boxes/Rack
제품 유형:
Side Access Freezer Rack (Adjustable)
용도(장비):
17.3, 23 cu. ft. Upright Freezers

Forma™ 900, 7000 Series and TSE/TSD Series Freezer용 Thermo Scientific™ Rack으로 초저온 냉동고의 보관 공간을 극대화하십시오. 슬라이딩 드로어 랙, 조절 가능한 측면 접근함 랙 및 표준형 또는 딥웰형의 마이크로플레이트 랙이 있습니다.

  • With sliding drawer racks
  • Adjustable side access box racks
  • Microplate racks in standard or deepwell formats
  • Warranty varies by country, please contact us for details
사양
깊이(영국식 단위)24.6 in.
깊이(미터법)62.5 cm
용도(장비)17.3, 23 cu. ft. Upright Freezers
높이(영국식 단위)11.6 in.
높이(미터법)29.5 cm
선반5
폭(영국식 단위)5.3 in.
폭(미터법)13.5 cm
오토클레이브 가능Autoclavable
색상Silver
보유Boxes (2 in.)
물질Stainless Steel
위치22 Boxes/Rack
제품 유형Side Access Freezer Rack (Adjustable)
형상Rectangular
적재 가능Yes
멸균Non-sterile
Unit SizeEach