Automation Tips for Caliper™ Zymark™ Liquid Handling Workstations
Thermo Scientific™
Automation Tips for Caliper™ Zymark™ Liquid Handling Workstations
Increase pipetting performance with these automation tips, which undergo a rigorous 15-point quality control program to ensure accuracy and consistency.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberVolume (Metric)FilteredSterilityFit for PurposeUnit Size
923-261200 μLNon-filteredSterileSterileCase of 4800
920-02150 μLFilteredSterileSterile and PCR ReadyCase of 4800
920-26150 μLNon-filteredSterileSterileCase of 4800
920-26250 μLNon-filteredNon-sterileCertified QualityCase of 4800
921-02180 μLFilteredSterileSterile and PCR ReadyCase of 4800
921-021PK
also known as 921-021
80 μLFilteredSterileSterile and PCR ReadyPack of 960
921-261100 μLNon-filteredSterileSterileCase of 4800
921-261PK
also known as 921-261
100 μLNon-filteredSterileSterilePack of 960
921-262100 μLNon-filteredNon-sterileCertified QualityCase of 4800
921-262PK
also known as 921-262
100 μLNon-filteredNon-sterileCertified QualityPack of 960
923-021150 μLFilteredSterileSterile and PCR ReadyCase of 4800
923-021PK
also known as 923-021
150 μLFilteredSterileSterile and PCR ReadyPack of 960
923-262200 μLNon-filteredNon-sterileCertified QualityCase of 4800
923-262PK
also known as 923-262
200 μLNon-filteredNon-sterileCertified QualityPack of 960
Catalog number 923-261
Price (USD)
245.00
Case of 4800
Volume (Metric):
200 μL
Filtered:
Non-filtered
Sterility:
Sterile
Fit for Purpose:
Sterile
Unit Size:
Case of 4800
Price (USD)
245.00
Case of 4800
Increase pipetting performance with Thermo Scientific™ Automation Tips for Caliper™ Zymark™ Liquid Handling Workstations, which undergo a rigorous 15-point quality control program to ensure consistency. Their superior tip straightness and a low coefficient of variation provide accurate and precise performance. Compatible with: Caliper/Zymark RapidPlate, Caliper Presto, Allegro, Zephyr and SciClone.
Features:
  • Available as sterile and filtered.
  • Sterile tips are certified free of RNase, DNase, DNA, endotoxin, bioburden and pyrogen.
  • Lot-tested during production on actual workstation for which they are created.
  • Low percentage coefficient of variation (%CV).
Warranty:A five (5)-year shelf life from the date of manufacture is recommended.
Specifications
ColorClear
FilteredNon-filtered
Fit for PurposeSterile
For Use With (Equipment)Zymark/Caliper Liquid Handling Workstations
FormatRack
Product LineCaliper Zymark
SterilitySterile
Tip StyleAutomation
Volume (Metric)200 μL
Unit SizeCase of 4800