Product Image
Thermo Scientific™
Automation Tips for PerkinElmer™/Packard™ Robotic Liquid Handling Systems with Varispan™ Dispense Arms
Pipette more effectively with Thermo Scientific™ Automation Tips for Perkin Elmer™/Packard™ Robotic Liquid Handling Systems with Varispan™ Dispense Arms.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberVolume (Metric)SterilityFiltered
927-021-0540 μLSterileFiltered
931-36230 μLNon-sterileFiltered
913-011130 μLSterileNon-filtered
929-262-05235 μLNon-sterileNon-filtered
925-262-0520 μLNon-sterileNon-filtered
929-261-05235 μLSterileNon-filtered
925-261-0520 μLSterileNon-filtered
925-021-0510 μLSterileFiltered
927-262-0550 μLNon-sterileNon-filtered
929-021-05180 μLSterileFiltered
Catalog number 927-021-05
Price (USD)
-
Volume (Metric):
40 μL
Sterility:
Sterile
Filtered:
Filtered
Pipette more effectively with Thermo Scientific™ Automation Tips for Perkin Elmer™/Packard™ Robotic Liquid Handling Systems with Varispan™ Dispense Arms, which undergo a rigorous 15-point quality control program to ensure consistency. This leads to accurate and precise performance due to superior tip straightness and a low coefficient of variation. The pipette tips are validated on the actual workstations for which they are created and the sterile pipette tips are certified free of RNase, DNase, DNA, endotoxin, bioburden and pyrogen.
  • Available in non-sterile, sterile, with Thermo Scientific™ ART™ filter technology and in low retention
  • Increased precision for the toughest multi-dispensing assays
  • Low percentage coefficient of variation (%CV)
Specifications
FilteredFiltered
For Use With (Equipment)Packard Liquid Handling System
FormatRack
SterilitySterile
Tip StyleLow Retention, Extended Length
Volume (Metric)40 μL
Unit SizeEach