Color-Rich Dyed Carboxylate-Modified Particles
Thermo Scientific™

Color-Rich Dyed Carboxylate-Modified Particles

Obtain superior test sensitivity in qualitative and quantitative lateral flow tests with Thermo Scientific™ Color-Rich Dyed Carboxylate-Modified Particles.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDiameter (Metric)QuantityDye Type
931018910202500.85 μm15 mLBlue
931018910203500.85 μm100 mLBlue
931018910204500.85 μm1000 mLBlue
DB1040CA0.4 μm15 mLBlue
DB1040CB0.4 μm100 mLBlue
DBK1040CA0.4 μm15 mLBlack
DBK1040CB0.4 μm100 mLBlack
DR1040CA0.4 μm15 mLRed
DR1040CB0.4 μm100 mLRed
Catalog number 93101891020250
Price (INR)
-
Diameter (Metric):
0.85 μm
Quantity:
15 mL
Dye Type:
Blue
Obtain superior test sensitivity in qualitative and quantitative lateral flow tests with Thermo Scientific™ Color-Rich Dyed Carboxylate-Modified Particles. Higher intensity dyed particles provide maximum color and brilliance, as well as immunologically reactive surfaces. These particles are composed of polystyrene with or without a copolymer grated surface for fast coupling and processing reactions. Available in blue, red or black internally dyed versions, these hydrophilic particles can bind ligands without any dye interference, feature high protein binding capacity; and have an optimized acid content.
These particles are internally dyed using the Thermo Scientific™ Color-Rich internal dyeing method or by utilizing the proprietary Firefli™ fluorescent dyeing process. Color-Rich dyeing methods provide exceptional color saturation, prevent dye leaching in aqueous media and leave the surface free for covalent coupling through a COOH group and optimal immunological reactivity.

Features:
  • Dye-free surface for coupling
  • High protein binding capacity
  • Hydrophilic—readily adsorbs and couples proteins
  • Optimizes assays by controlling sensitivity, specificity and stability
  • Optimized acid content
  • Easy one-step covalent coupling protocols
Benefits:
  • Ensures reproducibility
  • Binds ligands without dye interference
  • Offers the optimal size for most latex-based lateral flow tests: 0.4μm nominal diameter (DB1040CA through DBK1040CB)
  • Ensures uniform migration through membranes with a coefficient of variation (CV) in diameter of less than 5%

Recommended for:

  • Clinical diagnostics
  • Molecular biology
  • Lateral flow, turbidimetric and slide aggulation assays
Specifications
AdditiveNone
Binding PropertyCarboxylate-modified
Concentration2.5% solids
ContentsDyed polystyrene microspheres in water
Diameter (Metric) Mean0.85 μm
Diameter (Metric) Nominal0.85 μm
Dye TypeBlue
IncludesCertificate of Analysis
Parking Area100A²/group
Particle Density1.06g/cm³
Quantity15 mL
Refractive Index1.59 at 589nm (25°C)
StabilityProduct stable for a minimum of 24 months
Surface ModificationCarboxylate-Modified
Uniformity<5% CV
ColorBlue
Diameter (Metric)0.85 μm
MaterialPolystyrene or Polystyrene with Coploymer Grafted Surface
Product TypeCarboxylate-Modified Particle
Unit SizeEach
Contents & Storage
Unless otherwise stated, refrigerate (2-8°C) product when not in use but do not freeze. Store upright and keep bottle tightly sealed. Keep in original amber bottle and do not expose to light which can deteriorate the product. Mix product with gentle inversion by hand or vortex mixer.